The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135919.1 | NP_001129391.1 | 4 - 5 | 1686 | 88 |
NM_181785.3 | NP_861450.1 | 4 - 5 | 1686 | 88 | |
XM_005266361.2 | XP_005266418.1 | 4 - 5 | 1364 | 88 | |
GenBank mRNA | AF315594.1 | - | 4 - 5 | 1270 | 88 |
AK093932.1 | - | 3 - 4 | 2114 | 88 | |
AK098426.1 | - | 2 - 3 | 716 | 88 | |
AK124604.1 | - | 2 - 3 | 2379 | 88 | |
BC036662.1 | - | 4 - 5 | 1648 | 88 | |
BC068556.1 | - | 4 - 5 | 1514 | 88 | |
BX537565.1 | - | 4 - 5 | 1497 | 88 | |
CB851220.1 | - | 2 - 3 | 213 | 88 |