Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100406896-100407400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
UniGene
Hs.105191
Gene Chromosome Location
Chr.7: 100400826-100429239 on Build GRCh38
Transcripts
NM_001258008.1 NM_017984.4 XM_005250479.2 XM_005250480.2 XM_006716035.3 XM_006716036.2 XM_006716037.3 XM_006716038.3 XM_006716040.3 XM_017012379.1 XM_017012380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4614974 c.1041delT p.F347fs*14 Chr.7: 100407252-100407252 on Build GRCh38
4441276 c.1060C>A p.L354M Chr.7: 100407233-100407233 on Build GRCh38
5358374 c.1063C>T p.P355S Chr.7: 100407230-100407230 on Build GRCh38
1082839 c.1006G>T p.E336* Chr.7: 100407287-100407287 on Build GRCh38
3715648 c.1036C>G p.L346V Chr.7: 100407257-100407257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754435144 Chr.7: 100407209-100407209 on Build GRCh38
rs756956235 Chr.7: 100407350-100407350 on Build GRCh38
rs762724424 Chr.7: 100407181-100407181 on Build GRCh38
rs994620057 Chr.7: 100407029-100407029 on Build GRCh38
rs139991769 Chr.7: 100407073-100407073 on Build GRCh38
rs776825317 Chr.7: 100407264-100407264 on Build GRCh38
rs937551114 Chr.7: 100407233-100407233 on Build GRCh38
rs191329587 Chr.7: 100407062-100407062 on Build GRCh38
rs758142679 Chr.7: 100407298-100407298 on Build GRCh38
rs747110777 Chr.7: 100407258-100407258 on Build GRCh38
rs891998547 Chr.7: 100406951-100406951 on Build GRCh38
rs753677532 Chr.7: 100407344-100407344 on Build GRCh38
rs773951427 Chr.7: 100407312-100407312 on Build GRCh38
rs1053284000 Chr.7: 100406950-100406950 on Build GRCh38
rs1008993276 Chr.7: 100407315-100407315 on Build GRCh38
rs752219077 Chr.7: 100407246-100407246 on Build GRCh38
rs544426111 Chr.7: 100407054-100407054 on Build GRCh38
rs752670572 Chr.7: 100407214-100407214 on Build GRCh38
rs1000217 Chr.7: 100407147-100407147 on Build GRCh38
rs760560120 Chr.7: 100407077-100407077 on Build GRCh38
rs773693357 Chr.7: 100407290-100407290 on Build GRCh38
rs560654716 Chr.7: 100407202-100407202 on Build GRCh38
rs1040540317 Chr.7: 100407326-100407326 on Build GRCh38
rs770188603 Chr.7: 100407288-100407288 on Build GRCh38
rs768819366 Chr.7: 100407260-100407260 on Build GRCh38
rs182661375 Chr.7: 100407018-100407018 on Build GRCh38
rs904752156 Chr.7: 100406922-100406922 on Build GRCh38
rs900577727 Chr.7: 100407327-100407327 on Build GRCh38
rs766085914 Chr.7: 100407099-100407099 on Build GRCh38
rs76071385 Chr.7: 100406937-100406937 on Build GRCh38
rs992668350 Chr.7: 100407199-100407199 on Build GRCh38
rs560128327 Chr.7: 100407079-100407079 on Build GRCh38
rs964780757 Chr.7: 100406999-100406999 on Build GRCh38
rs780362839 Chr.7: 100407257-100407257 on Build GRCh38
rs748742165 Chr.7: 100407272-100407272 on Build GRCh38
rs527280132 Chr.7: 100407098-100407098 on Build GRCh38
rs536789722 Chr.7: 100407012-100407012 on Build GRCh38
rs866615773 Chr.7: 100406946-100406946 on Build GRCh38
rs984477078 Chr.7: 100407137-100407137 on Build GRCh38
rs754547851 Chr.7: 100407160-100407160 on Build GRCh38
rs780457069 Chr.7: 100407352-100407352 on Build GRCh38
rs746458994 Chr.7: 100407130-100407133 on Build GRCh38
rs977172914 Chr.7: 100407108-100407108 on Build GRCh38
rs200631169 Chr.7: 100407229-100407229 on Build GRCh38
rs766034230 Chr.7: 100407311-100407311 on Build GRCh38
rs186429031 Chr.7: 100407061-100407061 on Build GRCh38
rs996246493 Chr.7: 100407346-100407346 on Build GRCh38
rs1053741378 Chr.7: 100407249-100407249 on Build GRCh38
rs759492983 Chr.7: 100407228-100407228 on Build GRCh38
rs759155471 Chr.7: 100407323-100407323 on Build GRCh38
rs3900792 Chr.7: 100406954-100406954 on Build GRCh38
rs767062964 Chr.7: 100407324-100407324 on Build GRCh38
rs897519272 Chr.7: 100407317-100407317 on Build GRCh38
rs572330431 Chr.7: 100406996-100406996 on Build GRCh38
rs1000215 Chr.7: 100406920-100406920 on Build GRCh38
rs1027473677 Chr.7: 100407031-100407031 on Build GRCh38
rs187521454 Chr.7: 100407242-100407242 on Build GRCh38
rs751075043 Chr.7: 100407206-100407206 on Build GRCh38
rs994986445 Chr.7: 100407322-100407322 on Build GRCh38
rs767471992 Chr.7: 100407213-100407213 on Build GRCh38
rs1014646780 Chr.7: 100406971-100406971 on Build GRCh38
rs145418256 Chr.7: 100407244-100407244 on Build GRCh38
rs375933544 Chr.7: 100407342-100407342 on Build GRCh38
rs373716300 Chr.7: 100407341-100407341 on Build GRCh38
rs936265201 Chr.7: 100407247-100407247 on Build GRCh38
rs571618234 Chr.7: 100406952-100406952 on Build GRCh38
rs763163066 Chr.7: 100407293-100407293 on Build GRCh38
rs2286266 Chr.7: 100406989-100406989 on Build GRCh38
rs576557481 Chr.7: 100407050-100407050 on Build GRCh38

View Full Product Details