The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206999.1 | NP_001193928.1 | 28 - 29 | 3646 | 68 |
NM_007174.2 | NP_009105.1 | 27 - 28 | 3520 | 68 | |
XM_006719206.2 | XP_006719269.1 | 28 - 29 | 3673 | 68 | |
XM_011537783.1 | XP_011536085.1 | 28 - 29 | 3673 | 68 | |
XM_011537784.1 | XP_011536086.1 | 28 - 29 | 3673 | 68 | |
XM_011537785.1 | XP_011536087.1 | 28 - 29 | 3673 | 68 | |
XM_011537787.1 | XP_011536089.1 | 27 - 28 | 3547 | 68 | |
XM_011537788.1 | XP_011536090.1 | 27 - 28 | 3547 | 68 | |
XM_011537789.1 | XP_011536091.1 | 20 - 21 | 2808 | 68 | |
XM_011537790.2 | XP_011536092.1 | 19 - 20 | 2376 | 68 | |
XM_011537791.2 | XP_011536093.1 | 19 - 20 | 3361 | 68 | |
XM_011537792.2 | XP_011536094.1 | 19 - 20 | 3506 | 68 | |
XM_017018735.1 | XP_016874224.1 | 28 - 29 | 3673 | 68 | |
XM_017018736.1 | XP_016874225.1 | 28 - 29 | 3673 | 68 | |
XM_017018737.1 | XP_016874226.1 | 28 - 29 | 3673 | 68 | |
XM_017018738.1 | XP_016874227.1 | 17 - 18 | 2917 | 68 | |
XM_017018739.1 | XP_016874228.1 | 18 - 19 | 3782 | 68 | |
GenBank mRNA | AB023166.2 | - | 19 - 20 | 3506 | 68 |
AY257469.1 | - | 27 - 28 | 3517 | 68 | |
AY681966.1 | - | 28 - 29 | 3646 | 68 |