The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000899.4 | NP_000890.1 | 10 - 10 | 4040 | 100 |
NM_003994.5 | NP_003985.2 | 9 - 9 | 3956 | 100 | |
GenBank mRNA | AK025245.1 | - | 1 - 1 | 447 | 100 |
AK055903.1 | - | 1 - 1 | 1450 | 100 | |
CR749222.1 | - | 9 - 9 | 3788 | 100 |