Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location
Chr.3: 35638981 - 35798933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267617.1 NP_001254546.1 12 - 13 1312 77
NM_001267619.1 NP_001254548.1 12 - 13 1337 77
NM_016300.4 NP_057384.2 14 - 15 1491 77
XM_005264811.2 XP_005264868.1 14 - 15 1520 77
XM_006712943.2 XP_006713006.1 13 - 14 1418 77
XM_006712944.2 XP_006713007.1 12 - 13 1358 77
XM_017005576.1 XP_016861065.1 14 - 15 2064 77
XM_017005577.1 XP_016861066.1 14 - 15 1514 77
XM_017005578.1 XP_016861067.1 14 - 15 1490 77
XM_017005580.1 XP_016861069.1 15 - 16 2156 77
XM_017005582.1 XP_016861071.1 14 - 15 2061 77
XM_017005583.1 XP_016861072.1 14 - 15 1511 77
XM_017005585.1 XP_016861074.1 13 - 14 2004 77
XM_017005586.1 XP_016861075.1 13 - 14 1460 77
XM_017005587.1 XP_016861076.1 13 - 14 1623 77
XM_017005588.1 XP_016861077.1 14 - 15 2096 77
XM_017005589.1 XP_016861078.1 14 - 15 1546 77
XM_017005590.1 XP_016861079.1 14 - 15 13183 77
XM_017005592.1 XP_016861081.1 13 - 14 1962 77
XM_017005593.1 XP_016861082.1 13 - 14 1508 77
XM_017005594.1 XP_016861083.1 13 - 14 1412 77
XM_017005595.1 XP_016861084.1 14 - 15 2054 77
XM_017005597.1 XP_016861086.1 13 - 14 1959 77
XM_017005598.1 XP_016861087.1 14 - 15 2051 77
XM_017005602.1 XP_016861091.1 12 - 13 1902 77
XM_017005603.1 XP_016861092.1 12 - 13 1521 77
XM_017005604.1 XP_016861093.1 12 - 13 1352 77
XM_017005605.1 XP_016861094.1 13 - 14 1994 77
XM_017005606.1 XP_016861095.1 13 - 14 1444 77
XM_017005608.1 XP_016861097.1 15 - 16 2155 77
XM_017005609.1 XP_016861098.1 15 - 16 2187 77
XM_017005610.1 XP_016861099.1 12 - 13 1853 77
XM_017005611.1 XP_016861100.1 11 - 12 1793 77
XM_017005612.1 XP_016861101.1 12 - 13 1850 77
GenBank mRNA AK294484.1 - 12 - 13 1350 77
AL133109.1 - 6 - 7 641 77
BC031106.1 - 1 - 2 203 77
BC036399.1 - 12 - 13 1317 77
BC051828.1 - 14 - 15 1491 77

View Full Product Details