The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287005.1 | NP_001273934.1 | 5 - 6 | 729 | 67 |
NM_001287008.1 | NP_001273937.1 | 3 - 4 | 426 | 67 | |
NM_145006.3 | NP_659443.1 | 4 - 5 | 615 | 67 | |
XM_011518358.1 | XP_011516660.1 | 7 - 8 | 1809 | 67 | |
XM_011518359.1 | XP_011516661.1 | 6 - 7 | 1759 | 67 | |
XM_011518361.2 | XP_011516663.1 | 7 - 8 | 871 | 67 | |
XM_011518362.1 | XP_011516664.1 | 5 - 6 | 1645 | 67 | |
XM_017014448.1 | XP_016869937.1 | 5 - 6 | 916 | 67 | |
XM_017014449.1 | XP_016869938.1 | 4 - 5 | 797 | 67 | |
XM_017014450.1 | XP_016869939.1 | 6 - 7 | 756 | 67 | |
XM_017014451.1 | XP_016869940.1 | 4 - 5 | 1365 | 67 | |
GenBank mRNA | AI452970.1 | - | 3 - 4 | 335 | 67 |
AK128289.1 | - | 2 - 3 | 2583 | 67 | |
AL137623.1 | - | - | 165 | 67 | |
AY358190.1 | - | 5 - 6 | 657 | 67 | |
BC014601.2 | - | 4 - 5 | 596 | 67 | |
BC041834.1 | - | 2 - 3 | 2509 | 67 |