The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290184.1 | NP_001277113.1 | 3 - 4 | 452 | 69 |
NM_001290186.1 | NP_001277115.1 | 2 - 3 | 313 | 69 | |
NM_015331.2 | NP_056146.1 | 2 - 3 | 313 | 69 | |
XM_005245053.4 | XP_005245110.1 | 2 - 3 | 530 | 69 | |
XM_011509363.2 | XP_011507665.1 | 2 - 3 | 530 | 69 | |
GenBank mRNA | AF240468.1 | - | 2 - 3 | 331 | 69 |
AK299142.1 | - | 2 - 3 | 200 | 69 | |
AK310741.1 | - | 3 - 4 | 393 | 69 | |
AK314764.1 | - | 2 - 3 | 220 | 69 | |
AY359120.1 | - | 2 - 3 | 190 | 69 | |
BC047621.1 | - | 3 - 4 | 409 | 69 | |
D87442.1 | - | 2 - 3 | 187 | 69 |