Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
Chromosome Location
Chr.22: 19130279 - 19144726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022719.2 NP_073210.1 10 - 10 2116 67
NR_134304.1 10 - 10 2193 67
GenBank mRNA DB090071.1 - 1 - 1 415 67
L77565.1 - 1 - 1 151 67

View Full Product Details