Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27559878-27560393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
UniGene
Hs.493639
Gene Chromosome Location
Chr.9: 27546546-27573866 on Build GRCh38
Transcripts
NM_001256054.2 NM_018325.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1108023 c.672_682del11 p.S225fs*11 Chr.9: 27560283-27560293 on Build GRCh38
6256977 c.668C>A p.A223D Chr.9: 27560297-27560297 on Build GRCh38
6374310 c.719G>T p.S240I Chr.9: 27560246-27560246 on Build GRCh38
3906745 c.671T>G p.I224S Chr.9: 27560294-27560294 on Build GRCh38
6212338 c.708A>T p.V236V Chr.9: 27560257-27560257 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1026478163 Chr.9: 27560132-27560132 on Build GRCh38
rs781714755 Chr.9: 27559965-27559965 on Build GRCh38
rs1016175853 Chr.9: 27559990-27559990 on Build GRCh38
rs147034723 Chr.9: 27560283-27560283 on Build GRCh38
rs533717750 Chr.9: 27559911-27559911 on Build GRCh38
rs774358 Chr.9: 27559999-27559999 on Build GRCh38
rs764644027 Chr.9: 27560257-27560257 on Build GRCh38
rs539300734 Chr.9: 27560115-27560115 on Build GRCh38
rs998355293 Chr.9: 27560008-27560008 on Build GRCh38
rs775685558 Chr.9: 27560118-27560118 on Build GRCh38
rs74619849 Chr.9: 27560266-27560266 on Build GRCh38
rs751866003 Chr.9: 27560259-27560259 on Build GRCh38
rs199891950 Chr.9: 27560328-27560328 on Build GRCh38
rs750678579 Chr.9: 27560263-27560263 on Build GRCh38
rs1028555136 Chr.9: 27560017-27560017 on Build GRCh38
rs938480304 Chr.9: 27560114-27560114 on Build GRCh38
rs192772526 Chr.9: 27560113-27560113 on Build GRCh38
rs760132434 Chr.9: 27560245-27560245 on Build GRCh38
rs1011196541 Chr.9: 27560336-27560336 on Build GRCh38
rs765907344 Chr.9: 27560250-27560250 on Build GRCh38
rs749427700 Chr.9: 27560313-27560313 on Build GRCh38
rs534747606 Chr.9: 27560073-27560073 on Build GRCh38
rs1012846119 Chr.9: 27559957-27559957 on Build GRCh38
rs1041923163 Chr.9: 27559907-27559907 on Build GRCh38
rs147898406 Chr.9: 27560107-27560107 on Build GRCh38
rs557691780 Chr.9: 27560124-27560124 on Build GRCh38
rs894400618 Chr.9: 27559951-27559951 on Build GRCh38
rs747828926 Chr.9: 27560338-27560338 on Build GRCh38
rs113506887 Chr.9: 27560146-27560146 on Build GRCh38
rs530713200 Chr.9: 27559949-27559949 on Build GRCh38
rs111301744 Chr.9: 27560068-27560068 on Build GRCh38
rs548820893 Chr.9: 27559959-27559959 on Build GRCh38
rs1028522516 Chr.9: 27560301-27560301 on Build GRCh38
rs768696956 Chr.9: 27560318-27560318 on Build GRCh38
rs28526385 Chr.9: 27559939-27559939 on Build GRCh38
rs924363887 Chr.9: 27560111-27560111 on Build GRCh38
rs763431979 Chr.9: 27560256-27560256 on Build GRCh38
rs781214485 Chr.9: 27560262-27560262 on Build GRCh38
rs1004326155 Chr.9: 27559979-27559979 on Build GRCh38
rs555236783 Chr.9: 27560254-27560254 on Build GRCh38
rs139743072 Chr.9: 27560295-27560295 on Build GRCh38
rs559255138 Chr.9: 27559988-27559988 on Build GRCh38
rs771617024 Chr.9: 27560190-27560190 on Build GRCh38
rs968869092 Chr.9: 27560089-27560089 on Build GRCh38
rs190377082 Chr.9: 27559931-27559931 on Build GRCh38
rs184260800 Chr.9: 27560189-27560189 on Build GRCh38
rs964660890 Chr.9: 27560339-27560339 on Build GRCh38
rs768791752 Chr.9: 27560207-27560207 on Build GRCh38
rs1024670440 Chr.9: 27560025-27560025 on Build GRCh38
rs915799914 Chr.9: 27560168-27560168 on Build GRCh38
rs781149836 Chr.9: 27560018-27560018 on Build GRCh38
rs1021313724 Chr.9: 27560084-27560084 on Build GRCh38
rs1048860037 Chr.9: 27560229-27560229 on Build GRCh38
rs773991269 Chr.9: 27560182-27560182 on Build GRCh38
rs772918404 Chr.9: 27560191-27560191 on Build GRCh38

View Full Product Details