Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14843453-14843974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs778946615 Chr.X: 14843669-14843669 on Build GRCh38
rs1020168794 Chr.X: 14843727-14843727 on Build GRCh38
rs749699443 Chr.X: 14843895-14843895 on Build GRCh38
rs2375726 Chr.X: 14843553-14843553 on Build GRCh38
rs757610874 Chr.X: 14843903-14843903 on Build GRCh38
rs746293753 Chr.X: 14843750-14843750 on Build GRCh38
rs962222970 Chr.X: 14843846-14843846 on Build GRCh38
rs143131218 Chr.X: 14843695-14843695 on Build GRCh38
rs748392793 Chr.X: 14843708-14843708 on Build GRCh38
rs187611308 Chr.X: 14843534-14843534 on Build GRCh38
rs749578530 Chr.X: 14843745-14843745 on Build GRCh38
rs201070097 Chr.X: 14843918-14843918 on Build GRCh38
rs777812135 Chr.X: 14843882-14843882 on Build GRCh38
rs755077854 Chr.X: 14843680-14843680 on Build GRCh38
rs767721906 Chr.X: 14843854-14843854 on Build GRCh38
rs770685871 Chr.X: 14843593-14843593 on Build GRCh38
rs200325876 Chr.X: 14843706-14843706 on Build GRCh38
rs753509167 Chr.X: 14843643-14843643 on Build GRCh38
rs767194178 Chr.X: 14843626-14843626 on Build GRCh38
rs1025182355 Chr.X: 14843867-14843867 on Build GRCh38
rs190579053 Chr.X: 14843753-14843753 on Build GRCh38
rs140363445 Chr.X: 14843805-14843805 on Build GRCh38
rs35806617 Chr.X: 14843805-14843806 on Build GRCh38
rs773885877 Chr.X: 14843584-14843584 on Build GRCh38
rs749454699 Chr.X: 14843745-14843750 on Build GRCh38
rs935647794 Chr.X: 14843525-14843525 on Build GRCh38
rs750381270 Chr.X: 14843836-14843836 on Build GRCh38
rs763963394 Chr.X: 14843632-14843632 on Build GRCh38
rs981068174 Chr.X: 14843949-14843949 on Build GRCh38
rs776505971 Chr.X: 14843818-14843818 on Build GRCh38
rs765307016 Chr.X: 14843825-14843825 on Build GRCh38
rs780116069 Chr.X: 14843712-14843712 on Build GRCh38
rs1034395123 Chr.X: 14843762-14843762 on Build GRCh38
rs759020755 Chr.X: 14843594-14843594 on Build GRCh38
rs752657026 Chr.X: 14843871-14843871 on Build GRCh38
rs756392097 Chr.X: 14843880-14843880 on Build GRCh38
rs769023794 Chr.X: 14843528-14843528 on Build GRCh38
rs763533295 Chr.X: 14843566-14843566 on Build GRCh38
rs755167575 Chr.X: 14843844-14843844 on Build GRCh38
rs757677526 Chr.X: 14843488-14843488 on Build GRCh38
rs150377883 Chr.X: 14843808-14843808 on Build GRCh38
rs760440816 Chr.X: 14843630-14843630 on Build GRCh38
rs755704953 Chr.X: 14843496-14843496 on Build GRCh38
rs927015579 Chr.X: 14843509-14843509 on Build GRCh38
rs778731279 Chr.X: 14843756-14843756 on Build GRCh38
rs948292480 Chr.X: 14843493-14843493 on Build GRCh38
rs1006842387 Chr.X: 14843678-14843678 on Build GRCh38
rs757248077 Chr.X: 14843651-14843651 on Build GRCh38
rs148257882 Chr.X: 14843736-14843736 on Build GRCh38
rs373465821 Chr.X: 14843662-14843662 on Build GRCh38
rs771958941 Chr.X: 14843925-14843925 on Build GRCh38
rs745355429 Chr.X: 14843559-14843564 on Build GRCh38
rs143434225 Chr.X: 14843501-14843501 on Build GRCh38
rs995483698 Chr.X: 14843709-14843709 on Build GRCh38
rs747865842 Chr.X: 14843919-14843919 on Build GRCh38
rs752337343 Chr.X: 14843627-14843627 on Build GRCh38
rs368933083 Chr.X: 14843752-14843752 on Build GRCh38
rs377742035 Chr.X: 14843769-14843769 on Build GRCh38
rs773654012 Chr.X: 14843564-14843564 on Build GRCh38
rs771278010 Chr.X: 14843572-14843572 on Build GRCh38
rs201436396 Chr.X: 14843670-14843670 on Build GRCh38
rs779449906 Chr.X: 14843909-14843909 on Build GRCh38
rs746453967 Chr.X: 14843929-14843929 on Build GRCh38
rs761492600 Chr.X: 14843820-14843820 on Build GRCh38

View Full Product Details