Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107628345-107628852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPS1
Entrez Gene ID
5631
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
UniGene
Hs.56
Gene Chromosome Location
Chr.X: 107628424-107651026 on Build GRCh38
Transcripts
NM_001204402.1 NM_002764.3

SNP Detail

SNP ID Chromosome Location
rs886554805 Chr.X: 107628631-107628631 on Build GRCh38
rs1047645641 Chr.X: 107628806-107628807 on Build GRCh38
rs750915295 Chr.X: 107628430-107628430 on Build GRCh38
rs756853882 Chr.X: 107628688-107628688 on Build GRCh38
rs375500956 Chr.X: 107628774-107628774 on Build GRCh38
rs756583830 Chr.X: 107628766-107628766 on Build GRCh38
rs193273254 Chr.X: 107628787-107628787 on Build GRCh38
rs751085506 Chr.X: 107628682-107628682 on Build GRCh38
rs752590804 Chr.X: 107628640-107628640 on Build GRCh38
rs199968422 Chr.X: 107628556-107628556 on Build GRCh38
rs916698071 Chr.X: 107628534-107628534 on Build GRCh38
rs747701318 Chr.X: 107628806-107628806 on Build GRCh38
rs989529177 Chr.X: 107628526-107628526 on Build GRCh38
rs758827912 Chr.X: 107628454-107628454 on Build GRCh38
rs750791916 Chr.X: 107628763-107628763 on Build GRCh38
rs748321395 Chr.X: 107628796-107628796 on Build GRCh38
rs920387457 Chr.X: 107628539-107628539 on Build GRCh38
rs367842963 Chr.X: 107628780-107628780 on Build GRCh38
rs780954680 Chr.X: 107628735-107628735 on Build GRCh38
rs761523210 Chr.X: 107628585-107628585 on Build GRCh38
rs778859413 Chr.X: 107628788-107628788 on Build GRCh38
rs768856537 Chr.X: 107628476-107628476 on Build GRCh38
rs1047795093 Chr.X: 107628624-107628624 on Build GRCh38
rs371006312 Chr.X: 107628593-107628593 on Build GRCh38
rs991150282 Chr.X: 107628506-107628506 on Build GRCh38
rs754682838 Chr.X: 107628652-107628652 on Build GRCh38
rs869025594 Chr.X: 107628674-107628674 on Build GRCh38
rs944685055 Chr.X: 107628633-107628633 on Build GRCh38
rs958026624 Chr.X: 107628510-107628510 on Build GRCh38
rs978379537 Chr.X: 107628384-107628384 on Build GRCh38
rs948531147 Chr.X: 107628541-107628541 on Build GRCh38
rs772406932 Chr.X: 107628579-107628579 on Build GRCh38
rs776399830 Chr.X: 107628596-107628596 on Build GRCh38
rs751262798 Chr.X: 107628553-107628553 on Build GRCh38
rs763453976 Chr.X: 107628658-107628658 on Build GRCh38
rs937911805 Chr.X: 107628460-107628460 on Build GRCh38
rs767417913 Chr.X: 107628750-107628751 on Build GRCh38
rs765129709 Chr.X: 107628625-107628625 on Build GRCh38

View Full Product Details