Product Details

Assay Reference Genome
Location

Chr.17:58214360 on build GRCh38
Cytoband
17q22
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
54903
Gene Symbol
MKS1
Gene Name
Meckel syndrome, type 1
Gene Aliases
BBS13, MES, MKS, POC12
Location
Chr.17:58205436-58219605 on build GRCh38
Assay Gene Location
Within Exon 7
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
MKS1 NM_001165927.1 6 607 NP_001159399.1
NM_001321268.1 5 520 NP_001308197.1
NM_001321269.1 6 618 NP_001308198.1
NM_017777.3 6 618 NP_060247.2
XM_005257485.3 6 599 XP_005257542.1
XM_006721965.2 5 452 XP_006722028.1
XM_011524957.2 6 598 XP_011523259.1
XM_011524958.2 6 598 XP_011523260.1
XM_011524959.2 6 597 XP_011523261.1
XM_011524960.2 6 597 XP_011523262.1
XM_017024803.1 6 566 XP_016880292.1
XM_017024804.1 6 566 XP_016880293.1
XM_017024805.1 6 632 XP_016880294.1
AK000352.1 3 288 BAA91105.1
AK301020.1 5 451
AK310815.1 6 607
BC010061.2 6 553 AAH10061.1
CR457229.1 2 114 CAG33510.1
DQ185029.1 6 618 AAZ94714.1
KU178661.1 2 114
KU178662.1 2 114
KU178663.1 2 114

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv517229 Chr17:58176226 - 58373163 on Build GRCh38 Gain+Loss TSPOAP1 SUPT4H1 MPO EPX LPO TSPOAP1-AS1 MKS1 RNF43 MIR4736 MIR142

View Full Product Details