Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 6011944-6012454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RA
Entrez Gene ID
3559
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
CD25, IDDM10, IL2R, IMD41, TCGFR, p55
UniGene
Hs.231367
Gene Chromosome Location
Chr.10: 6010694-6062370 on Build GRCh38
Transcripts
NM_000417.2 NM_001308242.1 NM_001308243.1

SNP Detail

SNP ID Chromosome Location
rs991426557 Chr.10: 6012204-6012204 on Build GRCh38
rs549930913 Chr.10: 6012086-6012086 on Build GRCh38
rs12719919 Chr.10: 6012195-6012195 on Build GRCh38
rs566194606 Chr.10: 6012196-6012196 on Build GRCh38
rs1036569161 Chr.10: 6012116-6012116 on Build GRCh38
rs886047076 Chr.10: 6011988-6011988 on Build GRCh38
rs760142892 Chr.10: 6011987-6011987 on Build GRCh38
rs12722716 Chr.10: 6011984-6011984 on Build GRCh38
rs992186765 Chr.10: 6011988-6011988 on Build GRCh38
rs553112485 Chr.10: 6012401-6012401 on Build GRCh38
rs576607759 Chr.10: 6012384-6012384 on Build GRCh38
rs56206280 Chr.10: 6012290-6012290 on Build GRCh38
rs907888953 Chr.10: 6012238-6012238 on Build GRCh38
rs960634389 Chr.10: 6012361-6012361 on Build GRCh38
rs76242406 Chr.10: 6012163-6012163 on Build GRCh38
rs181357994 Chr.10: 6012411-6012411 on Build GRCh38
rs541601070 Chr.10: 6012273-6012273 on Build GRCh38
rs931147337 Chr.10: 6012351-6012351 on Build GRCh38
rs12722601 Chr.10: 6012428-6012428 on Build GRCh38
rs982040171 Chr.10: 6012214-6012214 on Build GRCh38
rs949490384 Chr.10: 6012213-6012213 on Build GRCh38
rs984852514 Chr.10: 6012402-6012402 on Build GRCh38
rs1050038084 Chr.10: 6012355-6012355 on Build GRCh38
rs768574986 Chr.10: 6012419-6012419 on Build GRCh38
rs886047077 Chr.10: 6012200-6012200 on Build GRCh38
rs147669116 Chr.10: 6011983-6011983 on Build GRCh38
rs1019307368 Chr.10: 6012016-6012016 on Build GRCh38
rs909348135 Chr.10: 6012104-6012104 on Build GRCh38
rs188613092 Chr.10: 6012394-6012394 on Build GRCh38
rs948136085 Chr.10: 6012096-6012096 on Build GRCh38
rs933640701 Chr.10: 6012174-6012174 on Build GRCh38
rs903683997 Chr.10: 6012266-6012266 on Build GRCh38
rs916642780 Chr.10: 6012212-6012212 on Build GRCh38
rs531419003 Chr.10: 6012034-6012034 on Build GRCh38
rs41290329 Chr.10: 6012120-6012120 on Build GRCh38
rs764877940 Chr.10: 6012263-6012263 on Build GRCh38
rs985292993 Chr.10: 6012099-6012099 on Build GRCh38
rs75779793 Chr.10: 6012165-6012165 on Build GRCh38
rs1050695706 Chr.10: 6012193-6012193 on Build GRCh38
rs76471890 Chr.10: 6012166-6012166 on Build GRCh38
rs536972453 Chr.10: 6012267-6012267 on Build GRCh38
rs916647121 Chr.10: 6011995-6011995 on Build GRCh38
rs143416542 Chr.10: 6012105-6012105 on Build GRCh38
rs554828895 Chr.10: 6012356-6012356 on Build GRCh38
rs1023612429 Chr.10: 6012389-6012389 on Build GRCh38
rs909326184 Chr.10: 6012410-6012410 on Build GRCh38
rs898429956 Chr.10: 6012339-6012339 on Build GRCh38
rs1006609925 Chr.10: 6012260-6012260 on Build GRCh38

View Full Product Details