Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47468963-47469468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

SNP Detail

SNP ID Chromosome Location
rs947754207 Chr.11: 47469220-47469220 on Build GRCh38
rs530408082 Chr.11: 47469057-47469057 on Build GRCh38
rs999991162 Chr.11: 47469043-47469043 on Build GRCh38
rs1014823265 Chr.11: 47469410-47469410 on Build GRCh38
rs1024028481 Chr.11: 47469172-47469174 on Build GRCh38
rs1041173651 Chr.11: 47469346-47469346 on Build GRCh38
rs1019645524 Chr.11: 47469028-47469028 on Build GRCh38
rs1040456092 Chr.11: 47469260-47469260 on Build GRCh38
rs1026952873 Chr.11: 47469087-47469087 on Build GRCh38
rs899934764 Chr.11: 47469363-47469363 on Build GRCh38
rs767744332 Chr.11: 47469234-47469234 on Build GRCh38
rs749863564 Chr.11: 47469374-47469374 on Build GRCh38
rs1000510521 Chr.11: 47469360-47469360 on Build GRCh38
rs1015204721 Chr.11: 47469157-47469157 on Build GRCh38
rs182959427 Chr.11: 47469179-47469179 on Build GRCh38
rs903889012 Chr.11: 47469338-47469338 on Build GRCh38
rs532658738 Chr.11: 47469302-47469302 on Build GRCh38
rs901940249 Chr.11: 47469267-47469267 on Build GRCh38
rs985094255 Chr.11: 47469240-47469242 on Build GRCh38
rs758078034 Chr.11: 47469382-47469382 on Build GRCh38
rs74571537 Chr.11: 47469257-47469257 on Build GRCh38
rs993247781 Chr.11: 47469428-47469428 on Build GRCh38
rs961930086 Chr.11: 47469425-47469425 on Build GRCh38
rs779798076 Chr.11: 47469386-47469386 on Build GRCh38
rs750524281 Chr.11: 47469337-47469337 on Build GRCh38
rs548088351 Chr.11: 47469315-47469315 on Build GRCh38
rs1042413136 Chr.11: 47469317-47469317 on Build GRCh38
rs984721567 Chr.11: 47469134-47469134 on Build GRCh38
rs561614150 Chr.11: 47469014-47469014 on Build GRCh38
rs980856149 Chr.11: 47469238-47469238 on Build GRCh38
rs1033253248 Chr.11: 47469133-47469133 on Build GRCh38
rs1009587894 Chr.11: 47469009-47469009 on Build GRCh38
rs137858363 Chr.11: 47469392-47469392 on Build GRCh38
rs35726813 Chr.11: 47469262-47469263 on Build GRCh38
rs892633751 Chr.11: 47469012-47469012 on Build GRCh38
rs944192389 Chr.11: 47469345-47469345 on Build GRCh38
rs917688228 Chr.11: 47469218-47469218 on Build GRCh38
rs1023976004 Chr.11: 47469400-47469400 on Build GRCh38
rs568996386 Chr.11: 47469399-47469399 on Build GRCh38
rs996822077 Chr.11: 47469069-47469069 on Build GRCh38
rs996847295 Chr.11: 47469366-47469366 on Build GRCh38
rs1032955619 Chr.11: 47469372-47469372 on Build GRCh38
rs892665786 Chr.11: 47469306-47469306 on Build GRCh38
rs952058343 Chr.11: 47469114-47469114 on Build GRCh38
rs924935395 Chr.11: 47469252-47469252 on Build GRCh38
rs945323224 Chr.11: 47469313-47469313 on Build GRCh38
rs971008781 Chr.11: 47469213-47469213 on Build GRCh38
rs991930510 Chr.11: 47469211-47469211 on Build GRCh38

View Full Product Details