Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
Chromosome Location
Chr.6: 75084326 - 75206262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004370.5 NP_004361.3 66 - 66 9547 133
NM_080645.2 NP_542376.2 51 - 51 6055 133
GenBank mRNA AJ420501.1 - 11 - 11 1035 133
AK057081.1 - 1 - 1 148 133
U73778.1 - 66 - 66 9494 133

View Full Product Details