Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 49169074-49169584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1
Entrez Gene ID
2346
Gene Name
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 49145094-49208706 on Build GRCh38
Transcripts
NM_001014986.1 NM_001193471.1 NM_001193472.1 NM_001193473.1 NM_004476.1 XM_011519958.2 XM_017017432.1 XM_017017433.1 XM_017017434.1 XM_017017435.1 XM_017017436.1 XM_017017437.1 XM_017017438.1 XM_017017439.1 XM_017017440.1 XM_017017441.1 XM_017017442.1 XM_017017443.1 XM_017017444.1 XM_017017445.1 XM_017017446.1 XM_017017447.1 XM_017017448.1 XM_017017449.1 XM_017017450.1 XM_017017451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1354257 c.1311G>T p.E437D Chr.11: 49169256-49169256 on Build GRCh38
1354258 c.1309-1G>A p.? Chr.11: 49169259-49169259 on Build GRCh38
7158738 c.1309G>C p.E437Q Chr.11: 49169258-49169258 on Build GRCh38
4033407 c.1366A>G p.I456V Chr.11: 49169201-49169201 on Build GRCh38
6248180 c.1343C>A p.A448D Chr.11: 49169224-49169224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977842773 Chr.11: 49169313-49169313 on Build GRCh38
rs1036381425 Chr.11: 49169530-49169530 on Build GRCh38
rs375484734 Chr.11: 49169228-49169228 on Build GRCh38
rs770157136 Chr.11: 49169246-49169246 on Build GRCh38
rs778138281 Chr.11: 49169177-49169177 on Build GRCh38
rs567538979 Chr.11: 49169389-49169389 on Build GRCh38
rs558275792 Chr.11: 49169503-49169503 on Build GRCh38
rs551128233 Chr.11: 49169385-49169385 on Build GRCh38
rs531116079 Chr.11: 49169308-49169308 on Build GRCh38
rs766207616 Chr.11: 49169287-49169287 on Build GRCh38
rs779839853 Chr.11: 49169221-49169221 on Build GRCh38
rs760936047 Chr.11: 49169300-49169300 on Build GRCh38
rs544200254 Chr.11: 49169507-49169507 on Build GRCh38
rs965831193 Chr.11: 49169360-49169360 on Build GRCh38
rs756906152 Chr.11: 49169309-49169309 on Build GRCh38
rs74799577 Chr.11: 49169365-49169365 on Build GRCh38
rs923706770 Chr.11: 49169337-49169337 on Build GRCh38
rs536483835 Chr.11: 49169398-49169398 on Build GRCh38
rs202721 Chr.11: 49169439-49169439 on Build GRCh38
rs757391966 Chr.11: 49169185-49169185 on Build GRCh38
rs751292367 Chr.11: 49169272-49169272 on Build GRCh38
rs768558882 Chr.11: 49169233-49169233 on Build GRCh38
rs756581989 Chr.11: 49169173-49169173 on Build GRCh38
rs746046584 Chr.11: 49169207-49169207 on Build GRCh38
rs111982341 Chr.11: 49169108-49169108 on Build GRCh38
rs772444946 Chr.11: 49169208-49169208 on Build GRCh38
rs779171479 Chr.11: 49169186-49169186 on Build GRCh38
rs975922082 Chr.11: 49169373-49169373 on Build GRCh38
rs909200298 Chr.11: 49169500-49169500 on Build GRCh38
rs907299325 Chr.11: 49169551-49169551 on Build GRCh38
rs552628726 Chr.11: 49169435-49169435 on Build GRCh38
rs754270799 Chr.11: 49169303-49169303 on Build GRCh38
rs764569671 Chr.11: 49169302-49169302 on Build GRCh38
rs931914198 Chr.11: 49169471-49169471 on Build GRCh38
rs35210122 Chr.11: 49169113-49169114 on Build GRCh38
rs571898670 Chr.11: 49169256-49169256 on Build GRCh38
rs753020865 Chr.11: 49169152-49169152 on Build GRCh38
rs776416420 Chr.11: 49169234-49169234 on Build GRCh38
rs575448614 Chr.11: 49169506-49169506 on Build GRCh38
rs762538705 Chr.11: 49169265-49169265 on Build GRCh38
rs538620934 Chr.11: 49169450-49169450 on Build GRCh38
rs756052948 Chr.11: 49169278-49169283 on Build GRCh38
rs1004948362 Chr.11: 49169176-49169176 on Build GRCh38
rs907351901 Chr.11: 49169188-49169188 on Build GRCh38
rs1049471213 Chr.11: 49169496-49169496 on Build GRCh38
rs766028126 Chr.11: 49169267-49169267 on Build GRCh38
rs772733825 Chr.11: 49169258-49169258 on Build GRCh38
rs1002961642 Chr.11: 49169560-49169560 on Build GRCh38
rs761785897 Chr.11: 49169244-49169244 on Build GRCh38
rs754427659 Chr.11: 49169184-49169184 on Build GRCh38
rs753632399 Chr.11: 49169308-49169308 on Build GRCh38

View Full Product Details