Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62706085-62706602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSCL2
Entrez Gene ID
26580
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
Gene Aliases
GNG3LG, HMN5, PELD, SPG17
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62709619 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

SNP Detail

SNP ID Chromosome Location
rs989052559 Chr.11: 62706402-62706402 on Build GRCh38
rs143831637 Chr.11: 62706560-62706560 on Build GRCh38
rs530728276 Chr.11: 62706393-62706393 on Build GRCh38
rs976435161 Chr.11: 62706388-62706389 on Build GRCh38
rs943874712 Chr.11: 62706352-62706353 on Build GRCh38
rs1041392330 Chr.11: 62706274-62706274 on Build GRCh38
rs979528077 Chr.11: 62706288-62706288 on Build GRCh38
rs1026405780 Chr.11: 62706160-62706160 on Build GRCh38
rs889105439 Chr.11: 62706414-62706414 on Build GRCh38
rs914327881 Chr.11: 62706258-62706258 on Build GRCh38
rs780318707 Chr.11: 62706192-62706192 on Build GRCh38
rs550768589 Chr.11: 62706409-62706409 on Build GRCh38
rs1053007992 Chr.11: 62706243-62706243 on Build GRCh38
rs1048778193 Chr.11: 62706348-62706348 on Build GRCh38
rs929263061 Chr.11: 62706386-62706387 on Build GRCh38
rs148920656 Chr.11: 62706433-62706433 on Build GRCh38
rs993571427 Chr.11: 62706148-62706148 on Build GRCh38
rs984455218 Chr.11: 62706514-62706514 on Build GRCh38
rs533416404 Chr.11: 62706278-62706278 on Build GRCh38
rs997750957 Chr.11: 62706408-62706408 on Build GRCh38
rs117597269 Chr.11: 62706279-62706279 on Build GRCh38
rs935849033 Chr.11: 62706237-62706237 on Build GRCh38
rs922003672 Chr.11: 62706187-62706187 on Build GRCh38
rs570775889 Chr.11: 62706423-62706423 on Build GRCh38
rs569333658 Chr.11: 62706385-62706385 on Build GRCh38
rs1035925033 Chr.11: 62706401-62706401 on Build GRCh38
rs916119480 Chr.11: 62706198-62706198 on Build GRCh38
rs544356542 Chr.11: 62706255-62706255 on Build GRCh38
rs1040925578 Chr.11: 62706354-62706354 on Build GRCh38
rs868448911 Chr.11: 62706138-62706138 on Build GRCh38
rs905998261 Chr.11: 62706395-62706396 on Build GRCh38
rs913080316 Chr.11: 62706296-62706296 on Build GRCh38
rs982124974 Chr.11: 62706209-62706209 on Build GRCh38
rs949745101 Chr.11: 62706245-62706245 on Build GRCh38
rs555049906 Chr.11: 62706487-62706487 on Build GRCh38
rs1048379689 Chr.11: 62706478-62706478 on Build GRCh38
rs952230391 Chr.11: 62706129-62706129 on Build GRCh38
rs901808041 Chr.11: 62706277-62706277 on Build GRCh38
rs368869119 Chr.11: 62706141-62706142 on Build GRCh38
rs376261584 Chr.11: 62706366-62706366 on Build GRCh38
rs1023506876 Chr.11: 62706394-62706403 on Build GRCh38
rs34509585 Chr.11: 62706451-62706452 on Build GRCh38
rs1048807482 Chr.11: 62706261-62706261 on Build GRCh38
rs374173364 Chr.11: 62706270-62706270 on Build GRCh38
rs979943520 Chr.11: 62706216-62706216 on Build GRCh38
rs897092846 Chr.11: 62706370-62706370 on Build GRCh38
rs1005060866 Chr.11: 62706136-62706136 on Build GRCh38
rs1018927012 Chr.11: 62706126-62706126 on Build GRCh38
rs111363747 Chr.11: 62706244-62706244 on Build GRCh38
rs189232519 Chr.11: 62706446-62706446 on Build GRCh38
rs1023114617 Chr.11: 62706287-62706287 on Build GRCh38
rs905530694 Chr.11: 62706250-62706250 on Build GRCh38
rs529417327 Chr.11: 62706290-62706290 on Build GRCh38
rs937030190 Chr.11: 62706345-62706345 on Build GRCh38
rs574944387 Chr.11: 62706495-62706495 on Build GRCh38
rs1001313790 Chr.11: 62706416-62706416 on Build GRCh38
rs1003017678 Chr.11: 62706540-62706540 on Build GRCh38
rs775909465 Chr.11: 62706266-62706266 on Build GRCh38
rs557894475 Chr.11: 62706561-62706561 on Build GRCh38
rs970321804 Chr.11: 62706205-62706205 on Build GRCh38
rs1032421684 Chr.11: 62706420-62706420 on Build GRCh38
rs539704135 Chr.11: 62706432-62706432 on Build GRCh38
rs985651375 Chr.11: 62706335-62706335 on Build GRCh38
rs886048443 Chr.11: 62706237-62706237 on Build GRCh38
rs951561158 Chr.11: 62706130-62706130 on Build GRCh38
rs1032452661 Chr.11: 62706289-62706289 on Build GRCh38
rs541886188 Chr.11: 62706111-62706111 on Build GRCh38
rs949961004 Chr.11: 62706324-62706324 on Build GRCh38
rs575422877 Chr.11: 62706251-62706251 on Build GRCh38
rs374089937 Chr.11: 62706285-62706285 on Build GRCh38
rs1003456487 Chr.11: 62706400-62706400 on Build GRCh38
rs570009564 Chr.11: 62706478-62706478 on Build GRCh38
rs981781317 Chr.11: 62706328-62706328 on Build GRCh38
rs966300055 Chr.11: 62706276-62706276 on Build GRCh38
rs984288142 Chr.11: 62706142-62706142 on Build GRCh38
rs911048681 Chr.11: 62706350-62706350 on Build GRCh38
rs976147290 Chr.11: 62706159-62706159 on Build GRCh38
rs937018930 Chr.11: 62706509-62706509 on Build GRCh38
rs1044447729 Chr.11: 62706521-62706521 on Build GRCh38
rs1015126927 Chr.11: 62706273-62706273 on Build GRCh38
rs887567122 Chr.11: 62706352-62706352 on Build GRCh38
rs905963616 Chr.11: 62706539-62706539 on Build GRCh38
rs1047746878 Chr.11: 62706387-62706387 on Build GRCh38
rs975542792 Chr.11: 62706403-62706403 on Build GRCh38
rs117862461 Chr.11: 62706280-62706280 on Build GRCh38
rs112877243 Chr.11: 62706230-62706230 on Build GRCh38
rs926947536 Chr.11: 62706331-62706331 on Build GRCh38
rs535236799 Chr.11: 62706466-62706466 on Build GRCh38
rs1027781198 Chr.11: 62706112-62706112 on Build GRCh38
rs1045373211 Chr.11: 62706247-62706247 on Build GRCh38
rs549450153 Chr.11: 62706291-62706291 on Build GRCh38
rs1036454759 Chr.11: 62706394-62706394 on Build GRCh38
rs192881882 Chr.11: 62706204-62706204 on Build GRCh38
rs1020107584 Chr.11: 62706463-62706463 on Build GRCh38
rs779044161 Chr.11: 62706404-62706404 on Build GRCh38
rs965997716 Chr.11: 62706145-62706145 on Build GRCh38

View Full Product Details