Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16986337-16986824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607287 c.3320C>T p.A1107V Chr.1: 16986548-16986548 on Build GRCh38
899711 c.3352C>A p.L1118M Chr.1: 16986516-16986516 on Build GRCh38
6009105 c.3426C>T p.L1142L Chr.1: 16986338-16986338 on Build GRCh38
5844514 c.3361_3363delCTG p.L1121delL Chr.1: 16986505-16986507 on Build GRCh38
1687074 c.3217C>T p.R1073W Chr.1: 16986823-16986823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769086688 Chr.1: 16986391-16986391 on Build GRCh38
rs765499517 Chr.1: 16986450-16986450 on Build GRCh38
rs770500433 Chr.1: 16986404-16986404 on Build GRCh38
rs779674096 Chr.1: 16986757-16986757 on Build GRCh38
rs758287038 Chr.1: 16986749-16986750 on Build GRCh38
rs750573377 Chr.1: 16986536-16986536 on Build GRCh38
rs564769077 Chr.1: 16986673-16986673 on Build GRCh38
rs765405083 Chr.1: 16986535-16986535 on Build GRCh38
rs971771759 Chr.1: 16986378-16986378 on Build GRCh38
rs560436144 Chr.1: 16986714-16986714 on Build GRCh38
rs149826987 Chr.1: 16986666-16986666 on Build GRCh38
rs139088170 Chr.1: 16986372-16986372 on Build GRCh38
rs757860911 Chr.1: 16986675-16986675 on Build GRCh38
rs781661852 Chr.1: 16986553-16986553 on Build GRCh38
rs377253172 Chr.1: 16986610-16986610 on Build GRCh38
rs778282746 Chr.1: 16986672-16986672 on Build GRCh38
rs199661502 Chr.1: 16986753-16986760 on Build GRCh38
rs377380258 Chr.1: 16986753-16986764 on Build GRCh38
rs758763354 Chr.1: 16986456-16986456 on Build GRCh38
rs56146840 Chr.1: 16986788-16986788 on Build GRCh38
rs927365149 Chr.1: 16986519-16986519 on Build GRCh38
rs746169788 Chr.1: 16986361-16986361 on Build GRCh38
rs764025000 Chr.1: 16986513-16986513 on Build GRCh38
rs369142791 Chr.1: 16986749-16986749 on Build GRCh38
rs375453342 Chr.1: 16986512-16986512 on Build GRCh38
rs759241323 Chr.1: 16986410-16986410 on Build GRCh38
rs775304821 Chr.1: 16986503-16986503 on Build GRCh38
rs913156094 Chr.1: 16986448-16986448 on Build GRCh38
rs1008998614 Chr.1: 16986367-16986367 on Build GRCh38
rs761814059 Chr.1: 16986612-16986612 on Build GRCh38
rs774002040 Chr.1: 16986406-16986406 on Build GRCh38
rs747548795 Chr.1: 16986388-16986388 on Build GRCh38
rs867332959 Chr.1: 16986488-16986488 on Build GRCh38
rs373514205 Chr.1: 16986484-16986484 on Build GRCh38
rs763038059 Chr.1: 16986781-16986781 on Build GRCh38
rs778370632 Chr.1: 16986592-16986592 on Build GRCh38
rs974555954 Chr.1: 16986663-16986663 on Build GRCh38
rs770410981 Chr.1: 16986487-16986487 on Build GRCh38
rs145674970 Chr.1: 16986525-16986525 on Build GRCh38
rs746638178 Chr.1: 16986771-16986771 on Build GRCh38
rs1020189579 Chr.1: 16986717-16986717 on Build GRCh38
rs751916254 Chr.1: 16986547-16986547 on Build GRCh38
rs766561219 Chr.1: 16986653-16986653 on Build GRCh38
rs1018692903 Chr.1: 16986368-16986368 on Build GRCh38
rs975127788 Chr.1: 16986715-16986715 on Build GRCh38
rs143483351 Chr.1: 16986750-16986761 on Build GRCh38
rs771715671 Chr.1: 16986598-16986598 on Build GRCh38
rs370677966 Chr.1: 16986611-16986611 on Build GRCh38
rs375233425 Chr.1: 16986725-16986725 on Build GRCh38
rs773379849 Chr.1: 16986629-16986629 on Build GRCh38
rs773291692 Chr.1: 16986776-16986776 on Build GRCh38
rs760637978 Chr.1: 16986435-16986435 on Build GRCh38
rs976061203 Chr.1: 16986793-16986793 on Build GRCh38
rs758675397 Chr.1: 16986539-16986539 on Build GRCh38
rs745487724 Chr.1: 16986491-16986491 on Build GRCh38
rs769818426 Chr.1: 16986774-16986774 on Build GRCh38
rs369278996 Chr.1: 16986792-16986792 on Build GRCh38
rs748975167 Chr.1: 16986667-16986668 on Build GRCh38
rs775186212 Chr.1: 16986433-16986433 on Build GRCh38
rs912787928 Chr.1: 16986700-16986700 on Build GRCh38
rs974089234 Chr.1: 16986742-16986742 on Build GRCh38
rs373315947 Chr.1: 16986789-16986789 on Build GRCh38
rs116456892 Chr.1: 16986784-16986784 on Build GRCh38
rs376475762 Chr.1: 16986785-16986785 on Build GRCh38
rs112261486 Chr.1: 16986541-16986541 on Build GRCh38
rs748774220 Chr.1: 16986395-16986395 on Build GRCh38
rs540833122 Chr.1: 16986703-16986703 on Build GRCh38
rs1047701202 Chr.1: 16986764-16986764 on Build GRCh38
rs763960380 Chr.1: 16986436-16986436 on Build GRCh38
rs115985012 Chr.1: 16986526-16986526 on Build GRCh38
rs533404016 Chr.1: 16986701-16986701 on Build GRCh38
rs375485447 Chr.1: 16986453-16986453 on Build GRCh38
rs2076605 Chr.1: 16986662-16986662 on Build GRCh38
rs774016983 Chr.1: 16986674-16986675 on Build GRCh38
rs757418466 Chr.1: 16986447-16986447 on Build GRCh38
rs937351387 Chr.1: 16986734-16986734 on Build GRCh38
rs768364385 Chr.1: 16986668-16986668 on Build GRCh38
rs567315420 Chr.1: 16986795-16986795 on Build GRCh38
rs201756175 Chr.1: 16986554-16986554 on Build GRCh38
rs780624699 Chr.1: 16986382-16986382 on Build GRCh38
rs757851198 Chr.1: 16986794-16986794 on Build GRCh38
rs374766933 Chr.1: 16986454-16986454 on Build GRCh38
rs372810630 Chr.1: 16986570-16986570 on Build GRCh38
rs773914880 Chr.1: 16986490-16986490 on Build GRCh38
rs767292295 Chr.1: 16986419-16986419 on Build GRCh38
rs767739342 Chr.1: 16986667-16986667 on Build GRCh38
rs886045577 Chr.1: 16986567-16986567 on Build GRCh38
rs768397313 Chr.1: 16986772-16986772 on Build GRCh38
rs34585239 Chr.1: 16986546-16986546 on Build GRCh38
rs537780162 Chr.1: 16986445-16986445 on Build GRCh38
rs988703795 Chr.1: 16986411-16986411 on Build GRCh38
rs753107684 Chr.1: 16986669-16986669 on Build GRCh38
rs9435735 Chr.1: 16986719-16986719 on Build GRCh38
rs927333814 Chr.1: 16986726-16986726 on Build GRCh38
rs137853968 Chr.1: 16986615-16986615 on Build GRCh38
rs930131952 Chr.1: 16986762-16986762 on Build GRCh38
rs369718815 Chr.1: 16986773-16986773 on Build GRCh38
rs369705373 Chr.1: 16986468-16986468 on Build GRCh38
rs755468538 Chr.1: 16986477-16986477 on Build GRCh38
rs779310083 Chr.1: 16986356-16986356 on Build GRCh38
rs781551851 Chr.1: 16986483-16986483 on Build GRCh38
rs925808360 Chr.1: 16986405-16986405 on Build GRCh38
rs541932176 Chr.1: 16986597-16986597 on Build GRCh38
rs769904401 Chr.1: 16986628-16986628 on Build GRCh38
rs755307953 Chr.1: 16986548-16986548 on Build GRCh38
rs771759787 Chr.1: 16986501-16986501 on Build GRCh38
rs568476959 Chr.1: 16986392-16986392 on Build GRCh38
rs761031832 Chr.1: 16986790-16986790 on Build GRCh38
rs957202477 Chr.1: 16986674-16986674 on Build GRCh38
rs376397035 Chr.1: 16986791-16986791 on Build GRCh38
rs780437824 Chr.1: 16986459-16986459 on Build GRCh38
rs756650754 Chr.1: 16986571-16986571 on Build GRCh38
rs763160625 Chr.1: 16986647-16986647 on Build GRCh38
rs528997540 Chr.1: 16986718-16986718 on Build GRCh38

View Full Product Details