Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20644408-20644916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PINK1
Entrez Gene ID
65018
Gene Name
PTEN induced putative kinase 1
Gene Aliases
BRPK, PARK6
UniGene
Hs.389171
Gene Chromosome Location
Chr.1: 20633455-20651511 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

Gene Symbol
PINK1-AS
Entrez Gene ID
100861548
Gene Name
PINK1 antisense RNA
Gene Aliases
NAPINK1, PINK1-AS1, PINK1AS
UniGene
Hs.710259
Gene Chromosome Location
Chr.1: 20642657-20652193 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5845325 c.865C>T p.P289S Chr.1: 20644578-20644578 on Build GRCh38
6903328 c.906delC p.L303fs*16 Chr.1: 20644619-20644619 on Build GRCh38
4383303 c.781T>C p.S261P Chr.1: 20644494-20644494 on Build GRCh38
1338543 c.809C>A p.P270H Chr.1: 20644522-20644522 on Build GRCh38
6746698 c.826C>T p.R276W Chr.1: 20644539-20644539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763193153 Chr.1: 20644773-20644773 on Build GRCh38
rs865957749 Chr.1: 20644868-20644868 on Build GRCh38
rs749553312 Chr.1: 20644590-20644590 on Build GRCh38
rs942299212 Chr.1: 20644757-20644758 on Build GRCh38
rs368453193 Chr.1: 20644565-20644565 on Build GRCh38
rs61735932 Chr.1: 20644548-20644548 on Build GRCh38
rs530433531 Chr.1: 20644550-20644550 on Build GRCh38
rs771310554 Chr.1: 20644454-20644454 on Build GRCh38
rs765479907 Chr.1: 20644721-20644721 on Build GRCh38
rs1013964019 Chr.1: 20644894-20644894 on Build GRCh38
rs148144773 Chr.1: 20644571-20644571 on Build GRCh38
rs146691996 Chr.1: 20644570-20644570 on Build GRCh38
rs776183250 Chr.1: 20644616-20644616 on Build GRCh38
rs199769220 Chr.1: 20644456-20644456 on Build GRCh38
rs754928971 Chr.1: 20644575-20644575 on Build GRCh38
rs773266267 Chr.1: 20644445-20644445 on Build GRCh38
rs938442681 Chr.1: 20644588-20644588 on Build GRCh38
rs113092523 Chr.1: 20644564-20644564 on Build GRCh38
rs767951846 Chr.1: 20644463-20644463 on Build GRCh38
rs747994596 Chr.1: 20644520-20644520 on Build GRCh38
rs897830836 Chr.1: 20644777-20644777 on Build GRCh38
rs570833319 Chr.1: 20644615-20644615 on Build GRCh38
rs367788556 Chr.1: 20644475-20644475 on Build GRCh38
rs112600292 Chr.1: 20644626-20644626 on Build GRCh38
rs760616885 Chr.1: 20644715-20644715 on Build GRCh38
rs771598213 Chr.1: 20644710-20644710 on Build GRCh38
rs748526651 Chr.1: 20644690-20644690 on Build GRCh38
rs560751763 Chr.1: 20644720-20644720 on Build GRCh38
rs774841958 Chr.1: 20644463-20644464 on Build GRCh38
rs762673179 Chr.1: 20644628-20644628 on Build GRCh38
rs542584036 Chr.1: 20644719-20644719 on Build GRCh38
rs904329400 Chr.1: 20644875-20644875 on Build GRCh38
rs987741774 Chr.1: 20644748-20644748 on Build GRCh38
rs28940284 Chr.1: 20644526-20644526 on Build GRCh38
rs769077171 Chr.1: 20644688-20644688 on Build GRCh38
rs752762678 Chr.1: 20644757-20644757 on Build GRCh38
rs142866598 Chr.1: 20644652-20644652 on Build GRCh38
rs775988170 Chr.1: 20644559-20644559 on Build GRCh38
rs200036561 Chr.1: 20644557-20644557 on Build GRCh38
rs772706227 Chr.1: 20644605-20644605 on Build GRCh38
rs761085380 Chr.1: 20644561-20644561 on Build GRCh38
rs557503577 Chr.1: 20644566-20644566 on Build GRCh38
rs1024670343 Chr.1: 20644511-20644511 on Build GRCh38
rs575081128 Chr.1: 20644687-20644687 on Build GRCh38
rs1052357378 Chr.1: 20644646-20644646 on Build GRCh38
rs889237078 Chr.1: 20644841-20644841 on Build GRCh38
rs186399714 Chr.1: 20644880-20644880 on Build GRCh38
rs7349186 Chr.1: 20644627-20644627 on Build GRCh38
rs181675228 Chr.1: 20644582-20644582 on Build GRCh38
rs372498707 Chr.1: 20644692-20644692 on Build GRCh38
rs56200357 Chr.1: 20644649-20644649 on Build GRCh38
rs758997850 Chr.1: 20644707-20644707 on Build GRCh38
rs529832802 Chr.1: 20644805-20644805 on Build GRCh38
rs200191396 Chr.1: 20644634-20644634 on Build GRCh38
rs1051761437 Chr.1: 20644804-20644804 on Build GRCh38
rs139226733 Chr.1: 20644665-20644665 on Build GRCh38
rs200708848 Chr.1: 20644517-20644517 on Build GRCh38
rs761346571 Chr.1: 20644667-20644669 on Build GRCh38
rs201939439 Chr.1: 20644466-20644466 on Build GRCh38
rs772510148 Chr.1: 20644551-20644551 on Build GRCh38
rs997542525 Chr.1: 20644546-20644546 on Build GRCh38
rs964717973 Chr.1: 20644752-20644752 on Build GRCh38
rs779060308 Chr.1: 20644600-20644600 on Build GRCh38
rs372280083 Chr.1: 20644515-20644515 on Build GRCh38
rs145350650 Chr.1: 20644893-20644893 on Build GRCh38
rs368019255 Chr.1: 20644717-20644717 on Build GRCh38
rs777088052 Chr.1: 20644578-20644578 on Build GRCh38
rs980886088 Chr.1: 20644521-20644521 on Build GRCh38
rs369566143 Chr.1: 20644577-20644577 on Build GRCh38
rs777920274 Chr.1: 20644589-20644589 on Build GRCh38
rs536146282 Chr.1: 20644647-20644647 on Build GRCh38
rs142183624 Chr.1: 20644661-20644661 on Build GRCh38
rs548506734 Chr.1: 20644540-20644540 on Build GRCh38
rs375197494 Chr.1: 20644642-20644642 on Build GRCh38
rs1043177976 Chr.1: 20644586-20644586 on Build GRCh38
rs749951690 Chr.1: 20644484-20644484 on Build GRCh38
rs758604046 Chr.1: 20644676-20644676 on Build GRCh38
rs367882094 Chr.1: 20644750-20644750 on Build GRCh38
rs74315355 Chr.1: 20644639-20644639 on Build GRCh38
rs730882053 Chr.1: 20644512-20644512 on Build GRCh38
rs780971409 Chr.1: 20644519-20644519 on Build GRCh38
rs74315359 Chr.1: 20644651-20644651 on Build GRCh38
rs548115934 Chr.1: 20644850-20644850 on Build GRCh38
rs398124659 Chr.1: 20644636-20644636 on Build GRCh38
rs74315358 Chr.1: 20644549-20644549 on Build GRCh38
rs202128685 Chr.1: 20644648-20644648 on Build GRCh38
rs757969881 Chr.1: 20644506-20644506 on Build GRCh38
rs913799290 Chr.1: 20644834-20644834 on Build GRCh38
rs778009684 Chr.1: 20644539-20644539 on Build GRCh38
rs759951885 Chr.1: 20644464-20644465 on Build GRCh38
rs115998841 Chr.1: 20644744-20644744 on Build GRCh38
rs1013707184 Chr.1: 20644477-20644477 on Build GRCh38
rs562392619 Chr.1: 20644797-20644797 on Build GRCh38
rs775334886 Chr.1: 20644712-20644712 on Build GRCh38
rs962124411 Chr.1: 20644533-20644533 on Build GRCh38
rs140266850 Chr.1: 20644728-20644728 on Build GRCh38
rs762565299 Chr.1: 20644747-20644747 on Build GRCh38
rs373473255 Chr.1: 20644593-20644593 on Build GRCh38
rs773906022 Chr.1: 20644644-20644644 on Build GRCh38
rs775975653 Chr.1: 20644634-20644634 on Build GRCh38
rs200949139 Chr.1: 20644662-20644662 on Build GRCh38
rs745996363 Chr.1: 20644604-20644604 on Build GRCh38
rs747550235 Chr.1: 20644684-20644684 on Build GRCh38
rs992581926 Chr.1: 20644733-20644733 on Build GRCh38
rs772496826 Chr.1: 20644432-20644432 on Build GRCh38
rs1046310145 Chr.1: 20644851-20644851 on Build GRCh38
rs374057536 Chr.1: 20644760-20644760 on Build GRCh38
rs145990823 Chr.1: 20644655-20644655 on Build GRCh38
rs773923961 Chr.1: 20644697-20644697 on Build GRCh38
rs558363399 Chr.1: 20644508-20644508 on Build GRCh38
rs781630957 Chr.1: 20644689-20644689 on Build GRCh38
rs531719864 Chr.1: 20644762-20644762 on Build GRCh38
rs998792054 Chr.1: 20644879-20644879 on Build GRCh38
rs772688164 Chr.1: 20644525-20644526 on Build GRCh38
rs550203458 Chr.1: 20644791-20644791 on Build GRCh38
rs774968834 Chr.1: 20644455-20644455 on Build GRCh38
rs544457754 Chr.1: 20644536-20644536 on Build GRCh38
rs201940147 Chr.1: 20644618-20644618 on Build GRCh38
rs747359904 Chr.1: 20644617-20644617 on Build GRCh38

View Full Product Details