The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000689.4 | NP_000680.2 | 4 - 5 | 764 | 80 |
GenBank mRNA | AB209821.1 | - | 4 - 5 | 490 | 80 |
AF003341.1 | - | 4 - 5 | 447 | 80 | |
AK000118.1 | - | 4 - 5 | 491 | 80 | |
AK026641.1 | - | 4 - 5 | 491 | 80 | |
AK293176.1 | - | 4 - 5 | 457 | 80 | |
AY390731.1 | - | 4 - 5 | 447 | 80 | |
BC001505.2 | - | 4 - 5 | 472 | 80 | |
BG773210.1 | - | 4 - 5 | 506 | 80 | |
BT006921.1 | - | 4 - 5 | 447 | 80 | |
EU668318.1 | - | 4 - 5 | 491 | 80 | |
EU794589.1 | - | 4 - 5 | 447 | 80 | |
FJ224286.1 | - | 4 - 5 | 500 | 80 |