The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127487.1 | NP_001120959.1 | 25 - 26 | 4666 | 123 |
NM_001458.4 | NP_001449.3 | 25 - 26 | 4666 | 123 | |
GenBank mRNA | AB208865.1 | - | 2 - 3 | 308 | 123 |
AB371585.1 | - | 25 - 26 | 4686 | 123 | |
AF089841.1 | - | 25 - 26 | 4506 | 123 | |
AF146692.1 | - | 25 - 26 | 4669 | 123 | |
AJ012737.1 | - | 25 - 26 | 4475 | 123 |