The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001807.4 | NP_001798.2 | 3 - 4 | 366 | 86 |
GenBank mRNA | AF081673.1 | - | 2 - 3 | 281 | 86 |
BC042510.1 | - | 3 - 4 | 351 | 86 | |
M85201.1 | - | 3 - 4 | 358 | 86 | |
S79774.1 | - | 3 - 4 | 360 | 86 | |
X54457.1 | - | 3 - 4 | 431 | 86 |