The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190415.1 | NP_001177344.1 | 5 - 6 | 682 | 60 |
NM_005641.3 | NP_005632.1 | 5 - 6 | 1011 | 60 | |
NM_139315.2 | NP_647476.1 | 5 - 6 | 713 | 60 | |
NR_033792.1 | 5 - 6 | 562 | 60 | ||
XM_006716100.2 | XP_006716163.1 | 6 - 7 | 804 | 60 | |
XM_006716101.2 | XP_006716164.1 | 5 - 6 | 641 | 60 | |
XM_006716102.2 | XP_006716165.1 | 5 - 6 | 508 | 60 | |
XM_011516542.2 | XP_011514844.1 | 6 - 7 | 783 | 60 | |
XM_011516543.2 | XP_011514845.1 | 5 - 6 | 536 | 60 | |
XM_017012568.1 | XP_016868057.1 | 5 - 6 | 981 | 60 | |
GenBank mRNA | AK300005.1 | - | 5 - 6 | 665 | 60 |
AK314428.1 | - | 5 - 6 | 988 | 60 | |
AK315077.1 | - | 5 - 6 | 663 | 60 | |
BC018115.1 | - | 5 - 6 | 656 | 60 | |
CR627390.1 | - | 5 - 6 | 540 | 60 | |
L25444.1 | - | 5 - 6 | 983 | 60 | |
U31659.1 | - | 4 - 5 | 767 | 60 |