Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR10H2
Entrez Gene ID
26538
Gene Name
olfactory receptor family 10 subfamily H member 2
Gene Aliases
-
Chromosome Location
Chr.19: 15728024 - 15729052 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013939.2 NP_039227.1 1 - 1 570 107
GenBank mRNA BC069085.1 - 1 - 1 550 107
BC069457.1 - 1 - 1 550 107
BC112178.1 - 1 - 1 582 107
BC113647.1 - 1 - 1 570 107

View Full Product Details