The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002251.2 | NP_001002251.2 | 4 - 5 | 1264 | 66 |
NM_001002252.2 | NP_001002252.2 | 4 - 5 | 1231 | 66 | |
NM_001278378.1 | NP_001265307.1 | 4 - 5 | 1120 | 66 | |
NM_001278379.1 | NP_001265308.1 | 4 - 5 | 871 | 66 | |
NM_001278380.1 | NP_001265309.1 | 4 - 5 | 1397 | 66 | |
NM_016638.3 | NP_057722.3 | 4 - 5 | 1231 | 66 | |
NM_018694.3 | NP_061164.3 | 4 - 5 | 1288 | 66 | |
NR_103512.1 | 4 - 5 | 1059 | 66 | ||
GenBank mRNA | AB035384.1 | - | 4 - 5 | 596 | 66 |
AF267748.1 | - | 4 - 5 | 534 | 66 | |
AK027285.1 | - | 3 - 4 | 933 | 66 | |
BC001958.1 | - | 4 - 5 | 565 | 66 | |
BC015569.2 | - | 4 - 5 | 561 | 66 | |
BC015909.1 | - | 4 - 5 | 1288 | 66 | |
BC094839.1 | - | 4 - 5 | 998 | 66 | |
BQ643520.1 | - | 4 - 5 | 668 | 66 | |
CD672894.1 | - | 4 - 5 | 470 | 66 | |
DQ099385.1 | - | 3 - 4 | 423 | 66 | |
EU624490.1 | - | 4 - 5 | 1231 | 66 | |
EU624491.1 | - | 4 - 5 | 962 | 66 |