The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033136.3 | NP_149127.1 | 2 - 3 | 744 | 79 |
GenBank mRNA | DA228737.1 | - | 2 - 3 | 315 | 79 |
DA326831.1 | - | 2 - 3 | 402 | 79 | |
S67292.1 | - | 1 - 2 | 171 | 79 | |
X65779.1 | - | 1 - 2 | 181 | 79 |