The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128217.2 | NP_001121689.1 | 2 - 3 | 418 | 71 |
NM_001317898.1 | NP_001304827.1 | 2 - 3 | 418 | 71 | |
NM_001317900.1 | NP_001304829.1 | 2 - 3 | 418 | 71 | |
NM_021144.3 | NP_066967.3 | 1 - 2 | 178 | 71 | |
NM_033222.4 | NP_150091.2 | 2 - 3 | 421 | 71 | |
GenBank mRNA | AF063020.1 | - | 2 - 3 | 387 | 71 |
AF098482.1 | - | 1 - 2 | 154 | 71 | |
AF098483.1 | - | 1 - 2 | 101 | 71 | |
AF432220.1 | - | 2 - 3 | 369 | 71 | |
BC022971.1 | - | 2 - 3 | 389 | 71 | |
BC033817.1 | - | 2 - 3 | 391 | 71 | |
BC044568.1 | - | 2 - 3 | 392 | 71 | |
BC064135.1 | - | 2 - 3 | 365 | 71 | |
BQ186453.1 | - | 1 - 2 | 178 | 71 | |
DA117094.1 | - | 2 - 3 | 404 | 71 |