The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018928.2 | NP_061751.1 | 1 - 1 | 153 | 69 |
NM_032406.1 | NP_115782.1 | 1 - 1 | 153 | 69 | |
GenBank mRNA | AF152338.1 | - | 1 - 1 | 153 | 69 |
AF152525.1 | - | 1 - 1 | 153 | 69 | |
BC042650.1 | - | 1 - 1 | 245 | 69 | |
BC101035.1 | - | 1 - 1 | 153 | 69 | |
BC101036.1 | - | 1 - 1 | 150 | 69 | |
BC101038.2 | - | 1 - 1 | 153 | 69 |