Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PALM2
- Entrez Gene ID
-
114299
- Gene Name
- paralemmin 2
- Gene Aliases
-
AKAP2
- Chromosome Location
-
Chr.9: 109640788 - 109951476 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053016.5 |
NP_443749.5 |
6 - 7 |
686 |
105 |
GenBank mRNA |
AJ312216.1 |
- |
6 - 7 |
544 |
105 |
- Gene Symbol
- PALM2-AKAP2
- Entrez Gene ID
-
445815
- Gene Name
- PALM2-AKAP2 readthrough
- Gene Aliases
-
AKAP2
- Chromosome Location
-
Chr.9: 109780297 - 110172512 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ312217.1 |
- |
6 - 7 |
540 |
105 |
View Full Product Details