Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FN1
- Entrez Gene ID
-
2335
- Gene Name
- fibronectin 1
- Gene Aliases
-
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
- Chromosome Location
-
Chr.2: 215360440 - 215436167 on Build GRCh38
View Full Product Details