Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TECTB
Entrez Gene ID
6975
Gene Name
tectorin beta
Gene Aliases
-
Chromosome Location
Chr.10: 112283656 - 112305035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058222.2 NP_478129.1 1 - 2 155 71
XM_017016604.1 XP_016872093.1 1 - 2 149 71
GenBank mRNA AF312827.1 - 1 - 2 76 71
BC113497.1 - 1 - 2 155 71
BC113499.1 - 1 - 2 155 71

View Full Product Details