Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMK2B
Entrez Gene ID
816
Gene Name
calcium/calmodulin dependent protein kinase II beta
Gene Aliases
CAM2, CAMK2, CAMKB
Chromosome Location
Chr.7: 44217150 - 44325994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220.4 NP_001211.3 3 - 4 428 62
NM_001293170.1 NP_001280099.1 3 - 4 428 62
NM_172078.2 NP_742075.1 3 - 4 428 62
NM_172079.2 NP_742076.1 3 - 4 428 62
NM_172080.2 NP_742077.1 3 - 4 428 62
NM_172081.2 NP_742078.1 3 - 4 428 62
NM_172082.2 NP_742079.1 3 - 4 428 62
NM_172083.2 NP_742080.1 3 - 4 428 62
NM_172084.2 NP_742081.1 3 - 4 428 62
XM_005249862.3 XP_005249919.1 3 - 4 304 62
XM_005249864.3 XP_005249921.1 3 - 4 304 62
XM_006715781.2 XP_006715844.1 3 - 4 304 62
XM_006715784.2 XP_006715847.1 3 - 4 304 62
XM_011515547.1 XP_011513849.1 3 - 4 304 62
XM_011515549.1 XP_011513851.1 3 - 4 304 62
XM_011515550.1 XP_011513852.1 3 - 4 304 62
XM_011515551.1 XP_011513853.1 3 - 4 304 62
XM_011515552.1 XP_011513854.1 3 - 4 304 62
XM_011515553.1 XP_011513855.1 3 - 4 304 62
XM_011515554.1 XP_011513856.1 3 - 4 304 62
XM_011515555.1 XP_011513857.1 3 - 4 304 62
XM_011515556.1 XP_011513858.1 3 - 4 304 62
XM_011515557.1 XP_011513859.1 3 - 4 304 62
XM_011515558.2 XP_011513860.1 3 - 4 304 62
XM_011515559.1 XP_011513861.1 3 - 4 213 62
XM_017012660.1 XP_016868149.1 3 - 4 304 62
XM_017012661.1 XP_016868150.1 3 - 4 304 62
XM_017012662.1 XP_016868151.1 3 - 4 304 62
XM_017012663.1 XP_016868152.1 3 - 4 213 62
XM_017012664.1 XP_016868153.1 3 - 4 304 62
XM_017012665.1 XP_016868154.1 3 - 4 304 62
XM_017012666.1 XP_016868155.1 3 - 4 213 62
XM_017012667.1 XP_016868156.1 3 - 4 213 62
XM_017012668.1 XP_016868157.1 3 - 4 213 62
XM_017012669.1 XP_016868158.1 3 - 4 213 62
XM_017012670.1 XP_016868159.1 3 - 4 213 62
GenBank mRNA AF078803.1 - 3 - 4 221 62
AF081572.1 - 3 - 4 221 62
AF081924.1 - 3 - 4 221 62
AF083419.1 - 3 - 4 221 62
AF140350.1 - 3 - 4 221 62
AJ252236.1 - 3 - 4 305 62
AJ252237.1 - 3 - 4 228 62
AJ252238.1 - 3 - 4 218 62
AK222703.1 - 3 - 4 252 62
AK290148.1 - 3 - 4 293 62
AK315663.1 - 3 - 4 335 62
BC019070.2 - 3 - 4 422 62
BI550751.1 - 3 - 4 433 62
BX445032.2 - 3 - 4 237 62
DA069361.1 - 2 - 3 80 62
DB175099.1 - 3 - 4 206 62
U23460.1 - 3 - 4 293 62

View Full Product Details