The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015072.4 | NP_055887.3 | 27 - 28 | 3442 | 71 |
GenBank mRNA | AB023215.1 | - | 25 - 26 | 3002 | 71 |
AK309748.1 | - | 28 - 29 | 3427 | 71 | |
AL133586.1 | - | 2 - 3 | 323 | 71 | |
AL136808.1 | - | 13 - 14 | 1890 | 71 | |
AY237126.1 | - | 27 - 28 | 3372 | 71 | |
BC136472.1 | - | 27 - 28 | 3332 | 71 | |
BC136473.1 | - | 27 - 28 | 3332 | 71 | |
CR533554.1 | - | 12 - 13 | 1820 | 71 |