The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324436.1 | NP_001311365.1 | 6 - 7 | 824 | 86 |
NM_001324438.1 | NP_001311367.1 | 6 - 7 | 824 | 86 | |
NM_018622.6 | NP_061092.3 | 6 - 7 | 824 | 86 | |
NR_136893.1 | 6 - 7 | 880 | 86 | ||
XM_005247582.4 | XP_005247639.1 | 6 - 7 | 819 | 86 | |
XM_017006800.1 | XP_016862289.1 | 6 - 7 | 819 | 86 | |
GenBank mRNA | AF197937.1 | - | 6 - 7 | 762 | 86 |
AK024765.1 | - | 6 - 7 | 796 | 86 | |
BC014058.2 | - | 6 - 7 | 782 | 86 | |
CA417847.1 | - | 4 - 5 | 86 |