Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- INTS4
- Entrez Gene ID
-
92105
- Gene Name
- integrator complex subunit 4
- Gene Aliases
-
INT4, MST093
- Chromosome Location
-
Chr.11: 77874491 - 77994849 on Build GRCh38
- Gene Symbol
- INTS4P1
- Entrez Gene ID
-
285905
- Gene Name
- integrator complex subunit 4 pseudogene 1
- Gene Aliases
-
INTS4L1
- Chromosome Location
-
Chr.7: 65141037 - 65197161 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK057766.1 |
- |
5 - 6 |
756 |
78 |
- Gene Symbol
- INTS4P2
- Entrez Gene ID
-
644619
- Gene Name
- integrator complex subunit 4 pseudogene 2
- Gene Aliases
-
INTS4L2
- Chromosome Location
-
Chr.7: 65647854 - 65718688 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027392.2 |
|
5 - 6 |
|
78 |
GenBank mRNA |
BC111554.1 |
- |
5 - 6 |
|
78 |
View Full Product Details