The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138392.3 | NP_612401.2 | 6 - 7 | 449 | 60 |
XM_006723474.1 | XP_006723537.1 | 6 - 7 | 449 | 60 | |
XM_006723475.1 | XP_006723538.1 | 6 - 7 | 449 | 60 | |
XM_011527492.1 | XP_011525794.1 | 6 - 7 | 449 | 60 | |
XM_011527493.1 | XP_011525795.1 | 5 - 6 | 345 | 60 | |
XM_017027473.1 | XP_016882962.1 | 6 - 7 | 449 | 60 | |
XM_017027474.1 | XP_016882963.1 | 5 - 6 | 345 | 60 | |
GenBank mRNA | AF258553.1 | - | 6 - 7 | 448 | 60 |
AK075057.1 | - | 6 - 7 | 445 | 60 | |
AK123695.1 | - | 4 - 5 | 427 | 60 | |
AK300808.1 | - | 4 - 5 | 328 | 60 | |
AK300819.1 | - | 6 - 7 | 399 | 60 | |
AK312748.1 | - | 6 - 7 | 413 | 60 | |
BC022855.1 | - | 6 - 7 | 420 | 60 | |
DB038927.1 | - | 6 - 7 | 449 | 60 |