Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
Chromosome Location
Chr.16: 46657979 - 46689232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018206.5 NP_060676.2 11 - 12 1470 142
XM_005256045.3 XP_005256102.1 10 - 11 1355 142
XM_011523227.2 XP_011521529.1 11 - 12 2200 142
GenBank mRNA AF175265.1 - 11 - 12 1428 142
AF183418.1 - 10 - 11 1379 142
AF186382.1 - 11 - 12 1424 142
AF191298.2 - 11 - 12 1398 142
AK001614.1 - 11 - 12 1418 142
AK023650.1 - 7 - 8 981 142
AK025774.1 - - 3078 142
AK223218.1 - 11 - 12 1389 142
AL136888.1 - 11 - 12 1418 142
AL512769.1 - 4 - 5 475 142
BC002414.2 - 10 - 11 1377 142
BC010362.1 - 11 - 12 1391 142
BC093036.1 - 11 - 12 1416 142
BX647213.1 - 6 - 7 1760 142

View Full Product Details