The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017744.4 | NP_060214.2 | 3 - 4 | 750 | 143 |
NM_138727.3 | NP_620055.1 | 3 - 4 | 699 | 143 | |
NM_138728.2 | NP_620056.1 | 3 - 4 | 750 | 143 | |
NM_138729.3 | NP_620057.1 | 3 - 4 | 750 | 143 | |
XM_005270963.4 | XP_005271020.1 | 3 - 4 | 273 | 143 | |
XM_005270964.3 | XP_005271021.1 | 2 - 3 | 365 | 143 | |
XM_011541627.2 | XP_011539929.1 | 3 - 4 | 459 | 143 | |
XM_011541628.2 | XP_011539930.1 | 3 - 4 | 459 | 143 | |
XM_011541629.2 | XP_011539931.1 | 3 - 4 | 473 | 143 | |
XM_011541630.2 | XP_011539932.1 | 3 - 4 | 1169 | 143 | |
XM_011541631.2 | XP_011539933.1 | 3 - 4 | 297 | 143 | |
XM_017001534.1 | XP_016857023.1 | 3 - 4 | 459 | 143 | |
XM_017001535.1 | XP_016857024.1 | 3 - 4 | 459 | 143 | |
XM_017001536.1 | XP_016857025.1 | 2 - 3 | 365 | 143 | |
XM_017001538.1 | XP_016857027.1 | 2 - 3 | 240 | 143 | |
GenBank mRNA | AB081317.1 | - | 3 - 4 | 521 | 143 |
AB081318.1 | - | 3 - 4 | 470 | 143 | |
AB081319.1 | - | 3 - 4 | 521 | 143 | |
AB081320.1 | - | 3 - 4 | 521 | 143 | |
AK291042.1 | - | 3 - 4 | 451 | 143 | |
AK301869.1 | - | 3 - 4 | 451 | 143 | |
AK304004.1 | - | 3 - 4 | 408 | 143 | |
AK308727.1 | - | 2 - 3 | 374 | 143 | |
BC027898.1 | - | 2 - 3 | 352 | 143 | |
BC037830.1 | - | 3 - 4 | 1163 | 143 | |
BC047392.1 | - | 3 - 4 | 476 | 143 | |
BC065501.1 | - | 3 - 4 | 657 | 143 | |
BX537988.1 | - | 4 - 5 | 612 | 143 | |
BX640667.1 | - | 3 - 4 | 426 | 143 |