Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38413538-38414032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1
Entrez Gene ID
2260
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
UniGene
Hs.264887
Gene Chromosome Location
Chr.8: 38411138-38468834 on Build GRCh38
Transcripts
NM_001174063.1 NM_001174064.1 NM_001174065.1 NM_001174066.1 NM_001174067.1 NM_015850.3 NM_023105.2 NM_023106.2 NM_023110.2 XM_006716303.2 XM_006716304.1 XM_006716306.2 XM_006716307.1 XM_006716309.3 XM_006716310.2 XM_006716311.1 XM_006716312.1 XM_006716313.2 XM_006716314.1 XM_011544443.2 XM_011544444.1 XM_011544445.2 XM_011544446.2 XM_011544447.2 XM_011544448.1 XM_011544449.1 XM_011544450.2 XM_011544451.1 XM_017013219.1 XM_017013220.1 XM_017013221.1 XM_017013222.1 XM_017013223.1 XM_017013224.1 XM_017013225.1 XM_017013226.1 XM_017013227.1 XM_017013228.1 XM_017013229.1 XM_017013230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2960412 c.2173G>A p.D725N Chr.8: 38414007-38414007 on Build GRCh38
6976632 c.2342G>A p.G781E Chr.8: 38413725-38413725 on Build GRCh38
5819490 c.2314G>A p.D772N Chr.8: 38413753-38413753 on Build GRCh38
6564086 c.2196_2205del10 p.M732fs*26 Chr.8: 38414005-38414014 on Build GRCh38
4767562 c.2262+2T>C p.? Chr.8: 38413916-38413916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374586530 Chr.8: 38413837-38413837 on Build GRCh38
rs544882235 Chr.8: 38413877-38413877 on Build GRCh38
rs752744088 Chr.8: 38413816-38413816 on Build GRCh38
rs929334595 Chr.8: 38413927-38413927 on Build GRCh38
rs896378105 Chr.8: 38413880-38413880 on Build GRCh38
rs376538647 Chr.8: 38413865-38413865 on Build GRCh38
rs369782405 Chr.8: 38413939-38413939 on Build GRCh38
rs747737281 Chr.8: 38413915-38413915 on Build GRCh38
rs201490643 Chr.8: 38413932-38413932 on Build GRCh38
rs753288703 Chr.8: 38413680-38413680 on Build GRCh38
rs763543799 Chr.8: 38413612-38413612 on Build GRCh38
rs886189861 Chr.8: 38413864-38413864 on Build GRCh38
rs727505377 Chr.8: 38414001-38414001 on Build GRCh38
rs768736835 Chr.8: 38413636-38413636 on Build GRCh38
rs562534007 Chr.8: 38413814-38413814 on Build GRCh38
rs772265332 Chr.8: 38413990-38413990 on Build GRCh38
rs777850582 Chr.8: 38413828-38413828 on Build GRCh38
rs866523242 Chr.8: 38413562-38413562 on Build GRCh38
rs200408837 Chr.8: 38413736-38413736 on Build GRCh38
rs121909643 Chr.8: 38413918-38413918 on Build GRCh38
rs121909644 Chr.8: 38413795-38413795 on Build GRCh38
rs764873744 Chr.8: 38413758-38413758 on Build GRCh38
rs746834138 Chr.8: 38413839-38413839 on Build GRCh38
rs781148536 Chr.8: 38413640-38413640 on Build GRCh38
rs749184176 Chr.8: 38413658-38413658 on Build GRCh38
rs752125402 Chr.8: 38413606-38413606 on Build GRCh38
rs763550889 Chr.8: 38413878-38413878 on Build GRCh38
rs752866466 Chr.8: 38413902-38413902 on Build GRCh38
rs374473310 Chr.8: 38413943-38413943 on Build GRCh38
rs1056712623 Chr.8: 38413875-38413875 on Build GRCh38
rs746602135 Chr.8: 38413746-38413746 on Build GRCh38
rs760108543 Chr.8: 38413973-38413973 on Build GRCh38
rs774120844 Chr.8: 38413665-38413665 on Build GRCh38
rs988440153 Chr.8: 38413566-38413566 on Build GRCh38
rs574034006 Chr.8: 38413751-38413751 on Build GRCh38
rs770921947 Chr.8: 38413664-38413664 on Build GRCh38
rs758677681 Chr.8: 38413632-38413632 on Build GRCh38
rs371776128 Chr.8: 38413614-38413614 on Build GRCh38
rs747228916 Chr.8: 38413635-38413635 on Build GRCh38
rs759376422 Chr.8: 38413669-38413669 on Build GRCh38
rs758272119 Chr.8: 38413836-38413836 on Build GRCh38
rs914122698 Chr.8: 38413582-38413582 on Build GRCh38
rs1014179319 Chr.8: 38413646-38413646 on Build GRCh38
rs374507681 Chr.8: 38413673-38413673 on Build GRCh38
rs940500421 Chr.8: 38413866-38413866 on Build GRCh38
rs555706876 Chr.8: 38413737-38413737 on Build GRCh38
rs767698667 Chr.8: 38413726-38413727 on Build GRCh38
rs370903617 Chr.8: 38413827-38413827 on Build GRCh38
rs943502843 Chr.8: 38413883-38413883 on Build GRCh38
rs760882547 Chr.8: 38414009-38414009 on Build GRCh38
rs775161322 Chr.8: 38413740-38413740 on Build GRCh38
rs2956723 Chr.8: 38413792-38413792 on Build GRCh38
rs56234888 Chr.8: 38413783-38413783 on Build GRCh38
rs751937328 Chr.8: 38413788-38413788 on Build GRCh38
rs377620009 Chr.8: 38413698-38413698 on Build GRCh38
rs989394574 Chr.8: 38413720-38413720 on Build GRCh38
rs867532966 Chr.8: 38413738-38413738 on Build GRCh38
rs370615877 Chr.8: 38413754-38413754 on Build GRCh38
rs999936730 Chr.8: 38413781-38413781 on Build GRCh38
rs886062919 Chr.8: 38413815-38413815 on Build GRCh38
rs371517132 Chr.8: 38413906-38413906 on Build GRCh38
rs377149398 Chr.8: 38413747-38413747 on Build GRCh38
rs780308433 Chr.8: 38413697-38413697 on Build GRCh38
rs781328162 Chr.8: 38413714-38413714 on Build GRCh38
rs149979921 Chr.8: 38413797-38413797 on Build GRCh38
rs755160898 Chr.8: 38413704-38413704 on Build GRCh38
rs757848509 Chr.8: 38413615-38413615 on Build GRCh38
rs369652756 Chr.8: 38413715-38413715 on Build GRCh38
rs763600740 Chr.8: 38413679-38413679 on Build GRCh38
rs376173540 Chr.8: 38413799-38413799 on Build GRCh38
rs771680156 Chr.8: 38413671-38413671 on Build GRCh38
rs769375368 Chr.8: 38413872-38413872 on Build GRCh38
rs17182463 Chr.8: 38413633-38413633 on Build GRCh38
rs762400946 Chr.8: 38413682-38413682 on Build GRCh38
rs773808959 Chr.8: 38413760-38413760 on Build GRCh38
rs756341817 Chr.8: 38413905-38413905 on Build GRCh38
rs765285994 Chr.8: 38413907-38413908 on Build GRCh38
rs760659513 Chr.8: 38413888-38413888 on Build GRCh38
rs776189467 Chr.8: 38413753-38413753 on Build GRCh38
rs758339769 Chr.8: 38413911-38413911 on Build GRCh38
rs747827840 Chr.8: 38413871-38413871 on Build GRCh38
rs746786094 Chr.8: 38413921-38413921 on Build GRCh38
rs773330316 Chr.8: 38413832-38413836 on Build GRCh38
rs527606454 Chr.8: 38413938-38413938 on Build GRCh38
rs17176067 Chr.8: 38413882-38413882 on Build GRCh38
rs17182456 Chr.8: 38413645-38413645 on Build GRCh38
rs577661678 Chr.8: 38413847-38413847 on Build GRCh38
rs745746766 Chr.8: 38413829-38413829 on Build GRCh38
rs763571736 Chr.8: 38413766-38413766 on Build GRCh38
rs759186266 Chr.8: 38413592-38413592 on Build GRCh38
rs768329777 Chr.8: 38413846-38413846 on Build GRCh38
rs775835652 Chr.8: 38414008-38414008 on Build GRCh38
rs771439684 Chr.8: 38413951-38413951 on Build GRCh38
rs775166971 Chr.8: 38413672-38413672 on Build GRCh38
rs759860221 Chr.8: 38413894-38413894 on Build GRCh38
rs765900637 Chr.8: 38413690-38413690 on Build GRCh38
rs56341011 Chr.8: 38413948-38413948 on Build GRCh38
rs767953986 Chr.8: 38413900-38413900 on Build GRCh38
rs886062918 Chr.8: 38413558-38413558 on Build GRCh38
rs369397371 Chr.8: 38413595-38413595 on Build GRCh38
rs770834000 Chr.8: 38413730-38413730 on Build GRCh38
rs375101187 Chr.8: 38413626-38413626 on Build GRCh38
rs886062917 Chr.8: 38413557-38413557 on Build GRCh38
rs771672913 Chr.8: 38413922-38413922 on Build GRCh38
rs55906164 Chr.8: 38413739-38413739 on Build GRCh38
rs760270008 Chr.8: 38413611-38413611 on Build GRCh38
rs369776263 Chr.8: 38413933-38413933 on Build GRCh38
rs774683007 Chr.8: 38413972-38413972 on Build GRCh38
rs774758084 Chr.8: 38413890-38413890 on Build GRCh38
rs905873179 Chr.8: 38413912-38413912 on Build GRCh38
rs573344794 Chr.8: 38413691-38413691 on Build GRCh38

View Full Product Details