The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003544.2 | NP_003535.1 | 1 - 1 | 311 | 73 |
GenBank mRNA | BC069467.1 | - | 1 - 1 | 311 | 73 |