Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE1A
Entrez Gene ID
5136
Gene Name
phosphodiesterase 1A
Gene Aliases
CAM-PDE-1A, HCAM-1, HCAM1, HSPDE1A
Chromosome Location
Chr.2: 182140035 - 182523375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003683.2 NP_001003683.1 6 - 7 767 103
NM_001258312.1 NP_001245241.1 6 - 7 794 103
NM_001258313.1 NP_001245242.1 5 - 6 946 103
NM_001258314.1 NP_001245243.1 4 - 5 623 103
NM_005019.4 NP_005010.2 5 - 6 809 103
XM_011511323.2 XP_011509625.1 5 - 6 586 103
XM_011511324.2 XP_011509626.1 6 - 7 814 103
XM_011511325.2 XP_011509627.1 5 - 6 941 103
XM_011511326.2 XP_011509628.1 4 - 5 618 103
XM_017004294.1 XP_016859783.1 5 - 6 809 103
XM_017004295.1 XP_016859784.1 6 - 7 814 103
XM_017004296.1 XP_016859785.1 6 - 7 736 103
XM_017004297.1 XP_016859786.1 6 - 7 677 103
XM_017004298.1 XP_016859787.1 6 - 7 859 103
XM_017004299.1 XP_016859788.1 6 - 7 814 103
XM_017004300.1 XP_016859789.1 6 - 7 859 103
XM_017004301.1 XP_016859790.1 6 - 7 664 103
XM_017004302.1 XP_016859791.1 5 - 6 941 103
GenBank mRNA AJ401610.1 - 5 - 6 528 103
AK294239.1 - 6 - 7 729 103
AK295657.1 - 6 - 7 599 103
AK301720.1 - 4 - 5 606 103
AL110263.1 - 5 - 6 920 103
BC022480.1 - 5 - 6 777 103
U40370.1 - 6 - 7 660 103

View Full Product Details