The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258312.1 | NP_001245241.1 | 15 - 16 | 1901 | 106 |
NM_005019.4 | NP_005010.2 | 14 - 15 | 1916 | 106 | |
XM_017004295.1 | XP_016859784.1 | 15 - 16 | 1921 | 106 | |
XM_017004296.1 | XP_016859785.1 | 15 - 16 | 1843 | 106 | |
XM_017004297.1 | XP_016859786.1 | 15 - 16 | 1784 | 106 | |
XM_017004298.1 | XP_016859787.1 | 15 - 16 | 1966 | 106 | |
XM_017004302.1 | XP_016859791.1 | 14 - 15 | 2048 | 106 | |
GenBank mRNA | AK294239.1 | - | 15 - 16 | 1836 | 106 |
AK295657.1 | - | 15 - 16 | 1706 | 106 | |
BC022480.1 | - | 14 - 15 | 1884 | 106 | |
BG196993.1 | - | - | 106 |