The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168618.1 | NP_001162089.1 | 5 - 6 | 680 | 85 |
NM_020153.3 | NP_064538.3 | 6 - 7 | 833 | 85 | |
XM_011542905.2 | XP_011541207.1 | 6 - 7 | 3575 | 85 | |
XM_011542906.2 | XP_011541208.1 | 5 - 6 | 3417 | 85 | |
XM_017018017.1 | XP_016873506.1 | 5 - 6 | 572 | 85 | |
XM_017018018.1 | XP_016873507.1 | 4 - 5 | 543 | 85 | |
GenBank mRNA | AJ249980.1 | - | 5 - 6 | 607 | 85 |
AK025480.1 | - | 6 - 7 | 792 | 85 | |
AK289521.1 | - | 5 - 6 | 531 | 85 | |
AL136934.1 | - | 5 - 6 | 515 | 85 | |
BC011647.2 | - | 5 - 6 | 518 | 85 | |
BC012802.2 | - | 5 - 6 | 611 | 85 | |
BC022856.1 | - | 5 - 6 | 641 | 85 | |
CR457344.1 | - | 3 - 4 | 261 | 85 |