Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRIQ3
Entrez Gene ID
127255
Gene Name
leucine rich repeats and IQ motif containing 3
Gene Aliases
LRRC44
Chromosome Location
Chr.1: 74026015 - 74198201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105659.1 NP_001099129.1 1 - 2 190 83
NM_001322315.1 NP_001309244.1 1 - 2 190 83
XM_011540658.1 XP_011538960.1 1 - 2 204 83
XM_011540663.2 XP_011538965.1 1 - 2 204 83
XM_017000286.1 XP_016855775.1 1 - 2 204 83
XM_017000287.1 XP_016855776.1 1 - 2 204 83
XM_017000288.1 XP_016855777.1 1 - 2 204 83
GenBank mRNA AK131476.2 - 1 - 2 147 83
BC020778.1 - 1 - 2 150 83
BX647210.1 - 1 - 2 150 83

View Full Product Details