Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSMEM1
Entrez Gene ID
136263
Gene Name
serine rich single-pass membrane protein 1
Gene Aliases
C7orf45
Chromosome Location
Chr.7: 130205399 - 130216869 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145268.3 NP_660311.1 1 - 2 235 66
GenBank mRNA AK097635.1 - 1 - 2 221 66
BC017587.2 - 1 - 2 239 66

View Full Product Details