Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAGP
Entrez Gene ID
57228
Gene Name
small cell adhesion glycoprotein
Gene Aliases
hSMAGP
Chromosome Location
Chr.12: 51245349 - 51270418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033873.1 NP_001029045.1 1 - 2 155 146
GenBank mRNA BC008712.1 - 1 - 2 143 146
BC120965.1 - 1 - 2 131 146
BC120966.1 - 1 - 2 131 146
BG740791.1 - 1 - 2 175 146

View Full Product Details