Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRG1L
Entrez Gene ID
127262
Gene Name
tumor protein p63 regulated 1-like
Gene Aliases
FAM79A, h-mover, mover
Chromosome Location
Chr.1: 3624992 - 3630131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC019034.2 - 2 - 3 311 72

View Full Product Details