The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288821.1 | NP_001275750.1 | 4 - 5 | 761 | 69 |
NM_001288823.1 | NP_001275752.1 | 5 - 6 | 1073 | 69 | |
NM_058170.3 | NP_477518.2 | 4 - 5 | 1012 | 69 | |
XM_017000240.1 | XP_016855729.1 | 5 - 6 | 916 | 69 | |
GenBank mRNA | AF397392.2 | - | 4 - 5 | 654 | 69 |
AF397394.1 | - | 4 - 5 | 683 | 69 | |
AF397396.1 | - | 5 - 6 | 744 | 69 | |
AK095551.1 | - | 2 - 3 | 238 | 69 | |
AK095724.1 | - | 4 - 5 | 808 | 69 | |
AK300870.1 | - | 4 - 5 | 761 | 69 | |
AY358722.1 | - | 4 - 5 | 725 | 69 | |
BC022531.1 | - | 4 - 5 | 770 | 69 |