The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014839.1 | NP_001014839.1 | 3 - 4 | 297 | 67 |
NM_001014841.1 | NP_001014841.1 | 2 - 3 | 229 | 67 | |
NM_014284.2 | NP_055099.1 | 2 - 3 | 569 | 67 | |
GenBank mRNA | AB011179.1 | - | 2 - 3 | 177 | 67 |
AB018739.2 | - | 3 - 4 | 281 | 67 | |
AB018740.2 | - | 2 - 3 | 213 | 67 | |
AL536768.3 | - | 2 - 3 | 548 | 67 | |
BC024592.1 | - | 2 - 3 | 543 | 67 |