The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135195.1 | NP_001128667.1 | 3 - 4 | 706 | 113 |
NM_173596.2 | NP_775867.2 | 5 - 6 | 735 | 113 | |
XM_005268803.1 | XP_005268860.1 | 3 - 4 | 623 | 113 | |
XM_011538198.1 | XP_011536500.1 | 3 - 4 | 688 | 113 | |
XM_011538199.1 | XP_011536501.1 | 5 - 6 | 742 | 113 | |
XM_011538200.1 | XP_011536502.1 | 4 - 5 | 692 | 113 | |
XM_011538201.1 | XP_011536503.1 | 4 - 5 | 666 | 113 | |
XM_017019185.1 | XP_016874674.1 | 3 - 4 | 666 | 113 | |
XM_017019186.1 | XP_016874675.1 | 5 - 6 | 853 | 113 | |
XM_017019187.1 | XP_016874676.1 | 3 - 4 | 602 | 113 | |
GenBank mRNA | AK172768.1 | - | 3 - 4 | 669 | 113 |
AK298331.1 | - | 3 - 4 | 347 | 113 | |
AK313188.1 | - | 5 - 6 | 698 | 113 | |
BC027884.1 | - | 3 - 4 | 580 | 113 |